gRNA Design Details
- Target DNA Sequence
- GGTAAAGCGACATCATGAAG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 20667506 - 20667528 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF7
- Entrez Gene ID
-
151449
- Gene Name
- growth differentiation factor 7
- Gene Aliases
-
BMP12
- Chromosome Location
-
Chr.2: 20666664 - 20671490 on GRCh38
- UniGene ID
- Hs.447688
- Species
-
Human