gRNA Design Details

Target DNA Sequence
GGTAAAGCGACATCATGAAG
PAM Sequence
TGG
Target locus
Chr.2: 20667506 - 20667528 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 90.7/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GDF7
Entrez Gene ID
151449
Gene Name
growth differentiation factor 7
Gene Aliases
BMP12
Chromosome Location
Chr.2: 20666664 - 20671490 on GRCh38
UniGene ID
Hs.447688
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_182828.3 NP_878248.2 1