gRNA Design Details

Target DNA Sequence
GCGCGCGTTTTCTCTCCTTT
PAM Sequence
GGG
Target locus
Chr.19: 7916362 - 7916384 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 93.04/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TGFBR3L
Entrez Gene ID
100507588
Gene Name
transforming growth factor beta receptor 3 like
Gene Aliases
TGFR-3L
Chromosome Location
Chr.19: 7915211 - 7919097 on GRCh38
UniGene ID
Hs.646773
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001195259.1 NP_001182188.1 1
XM_011527610.2 XP_011525912.1 1
XM_011527611.2 XP_011525913.1 1
XM_011527612.2 XP_011525914.1 1
XM_011527613.2 XP_011525915.1 1
XM_011527614.2 XP_011525916.1 1