gRNA Design Details

Target DNA Sequence
GGGACAACAGCAATATCCTA
PAM Sequence
AGG
Target locus
Chr.17: 63657871 - 63657893 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 89.52/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
MAP3K3
Entrez Gene ID
4215
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
MAPKKK3, MEKK3
Chromosome Location
Chr.17: 63622209 - 63696310 on GRCh38
UniGene ID
Hs.29282
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_002401.3 NP_002392.2 5
NM_203351.1 NP_976226.1 6
XM_005257376.3 XP_005257433.1 6
XM_005257377.3 XP_005257434.1 5
XM_005257378.2 XP_005257435.1 6