gRNA Design Details
- Target DNA Sequence
- CGGAGCTACAATCACCTTCA
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 44388459 - 44388481 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF10
- Entrez Gene ID
-
2255
- Gene Name
- fibroblast growth factor 10
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 44301655 - 44389780 on GRCh38
- UniGene ID
- Hs.248049
- Species
-
Human