Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001126050.1 | NP_001119522.1 | 3 |
NM_001126051.1 | NP_001119523.1 | 3 | |
NM_001319186.1 | NP_001306115.1 | 3 | |
NM_001319187.1 | NP_001306116.1 | 3 | |
NM_001319188.1 | NP_001306117.1 | 4 | |
NM_004494.2 | NP_004485.1 | 3 | |
NR_135008.1 | 3 | ||
XM_011509453.2 | XP_011507755.2 | 3 | |
XM_011509454.2 | XP_011507756.1 | 3 | |
XM_011509455.2 | XP_011507757.1 | 3 |