gRNA Design Details
- Target DNA Sequence
- GAAAGCGAAGTACATTCCCG
- PAM Sequence
- CGG
- Target locus
-
Chr.12: 7695473 - 7695495 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF3
- Entrez Gene ID
-
9573
- Gene Name
- growth differentiation factor 3
- Gene Aliases
-
KFS3, MCOP7, MCOPCB6
- Chromosome Location
-
Chr.12: 7689785 - 7695764 on GRCh38
- UniGene ID
- Hs.86232
- Species
-
Human