gRNA Design Details
- Target DNA Sequence
- ACCTGTCTGCCATCAAAGAA
- PAM Sequence
- AGG
- Target locus
-
Chr.12: 7690616 - 7690638 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF3
- Entrez Gene ID
-
9573
- Gene Name
- growth differentiation factor 3
- Gene Aliases
-
KFS3, MCOP7, MCOPCB6
- Chromosome Location
-
Chr.12: 7689785 - 7695764 on GRCh38
- UniGene ID
- Hs.86232
- Species
-
Human