gRNA Design Details
- Target DNA Sequence
- ACCGATGGAAGTTGAGCACT
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 49213283 - 49213305 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igf2bp3
- Entrez Gene ID
-
140488
- Gene Name
- insulin-like growth factor 2 mRNA binding protein 3
- Gene Aliases
-
2610101N11Rik, AA522010, AL022933, AU045931, IMP-3, IMP3, Koc13, Neilsen, mimp3
- Chromosome Location
-
Chr.6: 49085218 - 49215009 on GRCm38
- UniGene ID
- Mm.281018
- Species
-
Mouse