gRNA Design Details
- Target DNA Sequence
- TGCGCAGCCTTAACCTGAGT
- PAM Sequence
- GGG
- Target locus
-
Chr.10: 47322868 - 47322890 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF2
- Entrez Gene ID
-
2658
- Gene Name
- growth differentiation factor 2
- Gene Aliases
-
BMP-9, BMP9, HHT5
- Chromosome Location
-
Chr.10: 47322490 - 47326270 on GRCh38
- UniGene ID
- Hs.279463
- Species
-
Human