gRNA Design Details

Target DNA Sequence
TGCGCAGCCTTAACCTGAGT
PAM Sequence
GGG
Target locus
Chr.10: 47322868 - 47322890 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.69/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GDF2
Entrez Gene ID
2658
Gene Name
growth differentiation factor 2
Gene Aliases
BMP-9, BMP9, HHT5
Chromosome Location
Chr.10: 47322490 - 47326270 on GRCh38
UniGene ID
Hs.279463
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_016204.2 NP_057288.1 1