gRNA Design Details

Target DNA Sequence
AAACTTGGACGATGATAACT
PAM Sequence
TGG
Target locus
Chr.19: 8057840 - 8057862 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.54/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CCL25
Entrez Gene ID
6370
Gene Name
C-C motif chemokine ligand 25
Gene Aliases
Ckb15, SCYA25, TECK
Chromosome Location
Chr.19: 8052318 - 8062663 on GRCh38
UniGene ID
Hs.310511
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001201359.1 NP_001188288.1 5
NM_005624.3 NP_005615.2 5
XM_011528177.2 XP_011526479.1 6
XM_017027119.1 XP_016882608.1 6
XM_017027120.1 XP_016882609.1 6
XM_017027121.1 XP_016882610.1 6
XM_017027122.1 XP_016882611.1 6