Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001958.3 | NP_001949.1 | 4 | 863 | |
GenBank mRNA | AB209064.1 | - | 2 | 3103 | |
AB451389.1 | - | 3 | 534 | ||
AK307536.1 | - | 4 | 631 | ||
BC000432.2 | - | 3 | 602 | 2819899 | |
BC018855.2 | - | 2 | 244 | ||
BC110409.1 | - | 4 | 832 | 5741243 | |
L10340.1 | - | 3 | 451 | ||
X70940.1 | - | 3 | 617 |