Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000197.1 | NP_000188.1 | 3 | 268 | |
XM_011518618.2 | XP_011516920.1 | 6 | 740 | ||
XM_011518619.2 | XP_011516921.1 | 5 | 578 | ||
XM_017014671.1 | XP_016870160.1 | 5 | 2652 | ||
XM_017014672.1 | XP_016870161.1 | 5 | 2576 | ||
XM_017014673.1 | XP_016870162.1 | 5 | 2653 | ||
XM_017014674.1 | XP_016870163.1 | 5 | 2655 | ||
XM_017014676.1 | XP_016870165.1 | 5 | 2654 | ||
XM_017014677.1 | XP_016870166.1 | 5 | 2657 | ||
GenBank mRNA | BC034281.1 | - | 3 | 252 | |
BT019371.1 | - | 3 | 220 | ||
CR541894.1 | - | 3 | 220 | ||
KU178130.1 | - | 3 | 220 | ||
KU178131.1 | - | 3 | 220 | ||
KU178132.1 | - | 3 | 220 | ||
KU178133.1 | - | 3 | 220 | ||
U05659.1 | - | 3 | 268 |