Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318758.1 | NP_001305687.1 | 8 | 1797 | |
NM_002504.5 | NP_002495.2 | 8 | 1797 | ||
NM_147134.3 | NP_667345.1 | 8 | 1797 | ||
GenBank mRNA | AF332009.1 | - | 8 | 1702 | |
AK225750.1 | - | 8 | 1670 | ||
AK291643.1 | - | 8 | 1687 | ||
AL832342.1 | - | 8 | 1680 | ||
BC006447.1 | - | 7 | 1294 | ||
BC012151.1 | - | 8 | 1667 | 4558442 | |
U15306.1 | - | 7 | 1593 |