Gene Transcripts

Gene Symbol
VLDLR
Entrez Gene ID
7436
Gene Name
very low density lipoprotein receptor
Gene Aliases
CAMRQ1, CARMQ1, CHRMQ1, VLDL-R, VLDLRCH
Chromosome Location
Chr. 9: 2621679 - 2656103 on Build GRCh38
UniGene ID
Hs.370422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018056.2 NP_001018066.1 11, 12 2112
NM_001322225.1 NP_001309154.1 10, 11 1989
NM_001322226.1 NP_001309155.1 10, 11 1989
NM_003383.4 NP_003374.3 11, 12 2112
GenBank mRNA AB208822.1 - 12, 13 1803
AK092381.1 - 9, 10 1379
AK095744.1 - 7, 8 1192
AK289527.1 - 3, 4 495
BC136562.1 - 11, 12 1833 9020552
BC142653.1 - 11, 12 1774 40148037
BC144245.1 - 11, 12 1833 9052765
D16493.1 - 11, 12 1766
D16494.1 - 11, 12 1697
L20470.1 - 11, 12 2087
L22431.1 - 11, 12 1809
S73849.1 - 11, 12 1781

View Full Product Details