Gene Transcripts

Gene Symbol
CLEC5A
Entrez Gene ID
23601
Gene Name
C-type lectin domain family 5 member A
Gene Aliases
CLECSF5, MDL-1, MDL1
Chromosome Location
Chr. 7: 141927356 - 141947009 on Build GRCh38
UniGene ID
Hs.446235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301167.1 NP_001288096.1 5 519
NM_013252.2 NP_037384.1 6 564
XM_011515995.2 XP_011514297.1 4 451
XM_017011915.1 XP_016867404.1 5 522
XM_017011916.1 XP_016867405.1 6 590
XM_017011917.1 XP_016867406.1 5 521
GenBank mRNA AF139768.1 - 6 515
AJ271684.1 - 6 564
AK292766.1 - 6 553
AK295200.1 - 5 456
AK307095.1 - 4 382
AK313703.1 - 6 553
BC093714.1 - 5 388 7939559
BC112099.1 - 5 388 8327567
BC113098.1 - 6 506 40076723
BC113099.1 - 5 437 40076725
BC143392.1 - 4 319

View Full Product Details