Gene Transcripts

Gene Symbol
LPAR5
Entrez Gene ID
57121
Gene Name
lysophosphatidic acid receptor 5
Gene Aliases
GPR92, GPR93, KPG_010, LPA5
Chromosome Location
Chr. 12: 6618835 - 6636131 on Build GRCh38
UniGene ID
Hs.155538
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142961.1 NP_001136433.1 2 611
NM_020400.5 NP_065133.1 2 771
GenBank mRNA AB041943.1 - 1 588
AJ272207.1 - 1 330
AK313127.1 - 2 585
BC033571.2 - 2 562 4297800
BC072394.1 - 2 761 5751923

View Full Product Details