Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297778.1 | NP_001284707.1 | 3 | 377 | |
NM_001297779.1 | NP_001284708.1 | 3 | 362 | ||
NM_022787.3 | NP_073624.2 | 3 | 362 | ||
XM_011541971.2 | XP_011540273.1 | 3 | 315 | ||
XM_017002107.1 | XP_016857596.1 | 3 | 515 | ||
XM_017002108.1 | XP_016857597.1 | 3 | 315 | ||
GenBank mRNA | AF312734.1 | - | 2 | 306 | |
AF314163.1 | - | 2 | 218 | ||
AF459819.1 | - | 2 | 274 | ||
AK026065.1 | - | 3 | 340 | ||
AK222920.1 | - | 3 | 340 | ||
AK315640.1 | - | 3 | 316 | ||
BC014943.1 | - | 3 | 361 | 4874147 | |
BG751629.1 | - | 3 | 362 | ||
BP309588.1 | - | 3 | 348 | ||
BP339298.1 | - | 3 | 326 | ||
BU948893.1 | - | 3 | 333 |