Gene Transcripts
- Gene Symbol
- NBEAL2
- Entrez Gene ID
- 23218
- Gene Name
- neurobeachin like 2
- Gene Aliases
- BDPLT4, GPS
- Chromosome Location
- Chr. 3: 46979683 - 47009704 on Build GRCh38
- UniGene ID
- Hs.437043
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015175.2 |
NP_055990.1
|
40, 41 |
6729 |
|
XM_006713072.3 |
XP_006713135.1
|
39, 40 |
6645 |
|
XM_011533533.2 |
XP_011531835.1
|
40, 41 |
6726 |
|
XM_017006010.1 |
XP_016861499.1
|
40, 41 |
6726 |
|
XM_017006011.1 |
XP_016861500.1
|
40, 41 |
6582 |
|
XM_017006012.1 |
XP_016861501.1
|
39, 40 |
6645 |
|
XM_017006013.1 |
XP_016861502.1
|
40, 41 |
6726 |
|
XM_017006014.1 |
XP_016861503.1
|
39, 40 |
6501 |
|
XM_017006015.1 |
XP_016861504.1
|
38, 39 |
6357 |
|
XM_017006016.1 |
XP_016861505.1
|
35, 36 |
7303 |
|
XM_017006017.1 |
XP_016861506.1
|
17, 18 |
3145 |
|
GenBank mRNA |
AB011112.3 |
- |
26, 27 |
4413 |
|
AK131104.1 |
- |
40, 41 |
6569 |
|
AK293387.1 |
- |
1, 2 |
100 |
|
AK301269.1 |
- |
5, 6 |
792 |
|
AY358455.1 |
- |
10, 11 |
1679 |
|
BC029944.1 |
- |
2, 3 |
150 |
|
BC060874.1 |
- |
8, 9 |
1207 |
|
View Full Product Details