Gene Transcripts
- Gene Symbol
- MRPL4
- Entrez Gene ID
- 51073
- Gene Name
- mitochondrial ribosomal protein L4
- Gene Aliases
- CGI-28, L4mt
- Chromosome Location
- Chr. 19: 10251964 - 10260060 on Build GRCh38
- UniGene ID
- Hs.279652
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015956.2 |
NP_057040.2
|
3, 4 |
572 |
|
NM_146387.1 |
NP_666499.1
|
4, 5 |
436 |
|
NM_146388.1 |
NP_666500.1
|
3, 4 |
572 |
|
XM_011528045.2 |
XP_011526347.1
|
3, 4 |
567 |
|
GenBank mRNA |
AB049635.1 |
- |
3, 4 |
287 |
|
AF132962.1 |
- |
3, 4 |
533 |
|
AY007152.1 |
- |
2, 3 |
225 |
|
BC000756.2 |
- |
3, 4 |
526 |
2819730 |
BC009858.2 |
- |
3, 4 |
313 |
3928448 |
BG325905.1 |
- |
3, 4 |
573 |
|
BG758801.1 |
- |
4, 5 |
499 |
|
BM550378.1 |
- |
4, 5 |
434 |
|
View Full Product Details