Gene Transcripts
- Gene Symbol
- LRRC75A-AS1
- Entrez Gene ID
- 125144
- Gene Name
- LRRC75A antisense RNA 1
- Gene Aliases
- C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
- Chromosome Location
- Chr. 17: 16438987 - 16470648 on Build GRCh38
- UniGene ID
- Hs.368934
- Species
- Homo sapiens
- Gene Symbol
- LRRC75A
- Entrez Gene ID
- 388341
- Gene Name
- leucine rich repeat containing 75A
- Gene Aliases
- C17orf76, FAM211A
- Chromosome Location
- Chr. 17: 16442005 - 16492191 on Build GRCh38
- UniGene ID
- Hs.25425
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833245.1 |
- |
3 |
2721 |
|
AL833260.1 |
- |
3 |
2721 |
|
View Full Product Details