Gene Transcripts

Gene Symbol
LEMD2
Entrez Gene ID
221496
Gene Name
LEM domain containing 2
Gene Aliases
CTRCT42, NET25, dJ482C21.1
Chromosome Location
Chr. 6: 33771213 - 33794274 on Build GRCh38
UniGene ID
Hs.444845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181336.3 NP_851853.1 1 673
XM_017010437.1 XP_016865926.1 1 5818
GenBank mRNA AK074959.1 - 1 726
BC051803.1 - 1 663 5240212

View Full Product Details