Gene Transcripts
- Gene Symbol
- DUSP6
- Entrez Gene ID
- 1848
- Gene Name
- dual specificity phosphatase 6
- Gene Aliases
- HH19, MKP3, PYST1
- Chromosome Location
- Chr. 12: 89347825 - 89352859 on Build GRCh38
- UniGene ID
- Hs.298654
- Species
- Homo sapiens
- Gene Symbol
- LOC100131490
- Entrez Gene ID
- 100131490
- Gene Name
- uncharacterized LOC100131490
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.718640
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124265.1 |
- |
1 |
714 |
|
View Full Product Details