Gene Transcripts
- Gene Symbol
- DYRK1A
- Entrez Gene ID
- 1859
- Gene Name
- dual specificity tyrosine phosphorylation regulated kinase 1A
- Gene Aliases
- DYRK, DYRK1, HP86, MNB, MNBH, MRD7
- Chromosome Location
- Chr. 21: 37365790 - 37515376 on Build GRCh38
- UniGene ID
- Hs.368240
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001396.3 |
NP_001387.2
|
5, 6 |
747 |
|
NM_101395.2 |
NP_567824.1
|
7, 8 |
921 |
|
NM_130436.2 |
NP_569120.1
|
5, 6 |
2115 |
|
NM_130438.2 |
NP_569122.1
|
5, 6 |
748 |
|
XM_006723976.3 |
XP_006724039.1
|
6, 7 |
819 |
|
XM_006723977.3 |
XP_006724040.1
|
6, 7 |
1144 |
|
XM_006723978.3 |
XP_006724041.1
|
6, 7 |
1873 |
|
XM_006723979.3 |
XP_006724042.1
|
6, 7 |
979 |
|
XM_011529482.2 |
XP_011527784.1
|
5, 6 |
841 |
|
XM_011529483.2 |
XP_011527785.1
|
6, 7 |
3687 |
|
XM_011529484.2 |
XP_011527786.1
|
5, 6 |
814 |
|
XM_011529485.2 |
XP_011527787.1
|
5, 6 |
1330 |
|
XM_017028284.1 |
XP_016883773.1
|
6, 7 |
778 |
|
XM_017028285.1 |
XP_016883774.1
|
6, 7 |
1024 |
|
XM_017028286.1 |
XP_016883775.1
|
8, 9 |
1021 |
|
GenBank mRNA |
AF108830.1 |
- |
7, 8 |
921 |
|
D85759.1 |
- |
5, 6 |
713 |
|
D86550.1 |
- |
5, 6 |
2118 |
|
U52373.1 |
- |
5, 6 |
747 |
|
U58496.1 |
- |
5, 6 |
707 |
|
View Full Product Details