Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017924.3 | NP_001017924.1 | 2 | 197 | |
NM_001284223.1 | NP_001271152.1 | 2 | 197 | ||
GenBank mRNA | BC140765.1 | - | 2 | 197 | |
BC144475.1 | - | 2 | 197 | 9053002 | |
BX648524.1 | - | 2 | 137 |