Gene Transcripts

Gene Symbol
MYBPC3
Entrez Gene ID
4607
Gene Name
myosin binding protein C, cardiac
Gene Aliases
CMD1MM, CMH4, FHC, LVNC10, MYBP-C
Chromosome Location
Chr. 11: 47331406 - 47352702 on Build GRCh38
UniGene ID
Hs.524906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000256.3 NP_000247.2 2 319
GenBank mRNA AK291569.1 - 2 319
AK297843.1 - 2 319
AU140784.1 - 2 319
BC136543.1 - 2 296 9020533
BC136546.1 - 2 296 9020536
BC142685.1 - 2 299 40148564
BC151211.1 - 2 302 40147517
EF560722.1 - 2 320
EU747718.1 - 2 319
EU747719.1 - 2 319

View Full Product Details