Gene Transcripts

Gene Symbol
APH1A
Entrez Gene ID
51107
Gene Name
aph-1 homolog A, gamma-secretase subunit
Gene Aliases
6530402N02Rik, APH-1, APH-1A, CGI-78
Chromosome Location
Chr. 1: 150265399 - 150282291 on Build GRCh38
UniGene ID
Hs.108408
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077628.2 NP_001071096.1 4 797
NM_001243771.1 NP_001230700.1 3 626
NM_001243772.1 NP_001230701.1 3 587
NM_016022.3 NP_057106.2 4 797
NR_045033.1 4 661
NR_045034.1 2 552
NR_045035.1 3 723
XM_017001417.1 XP_016856906.1 5 1209
GenBank mRNA AF151835.1 - 4 499
AF508787.1 - 4 455
AK027879.1 - 4 567
AK075295.1 - 4 516
AK297680.1 - 4 427
AK298832.1 - 3 394
AK298876.1 - 3 502
AK300640.1 - 3 431
AK307358.1 - 2 359
AK314490.1 - 4 567
AX775955.1 - 4 533
AX775957.1 - 4 587
AY113698.1 - 4 455
AY113699.1 - 4 455
AY358951.1 - 4 601
BC001230.2 - 4 567 3457593
BC008732.2 - 4 554 3162330
BC009501.2 - 4 588 4040230
BC015568.2 - 4 556 3356401
BC017699.1 - 4 533 4177580
BC020590.1 - 4 587 4420942

View Full Product Details