Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104773351-104773841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
UniGene
Hs.525622
Gene Chromosome Location
Chr.14: 104769349-104795743 on Build GRCh38
Transcripts
NM_001014431.1 NM_001014432.1 NM_005163.2 XM_005267401.1 XM_017021075.1 XM_017021076.1 XM_017021077.1 XM_017021078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6506510 c.718T>C p.S240P Chr.14: 104773565-104773565 on Build GRCh38
2026431 c.815A>G p.Y272C Chr.14: 104773468-104773468 on Build GRCh38
953788 c.852G>A p.K284K Chr.14: 104773356-104773356 on Build GRCh38
6977169 c.728G>A p.R243H Chr.14: 104773555-104773555 on Build GRCh38
6598010 c.855C>T p.D285D Chr.14: 104773353-104773353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776642823 Chr.14: 104773584-104773584 on Build GRCh38
rs1049068367 Chr.14: 104773706-104773706 on Build GRCh38
rs536885193 Chr.14: 104773631-104773631 on Build GRCh38
rs1021679826 Chr.14: 104773660-104773660 on Build GRCh38
rs986307131 Chr.14: 104773667-104773667 on Build GRCh38
rs201044857 Chr.14: 104773476-104773476 on Build GRCh38
rs765714250 Chr.14: 104773591-104773591 on Build GRCh38
rs61761197 Chr.14: 104773810-104773810 on Build GRCh38
rs536368121 Chr.14: 104773691-104773691 on Build GRCh38
rs915517218 Chr.14: 104773419-104773419 on Build GRCh38
rs3730329 Chr.14: 104773603-104773603 on Build GRCh38
rs3730330 Chr.14: 104773451-104773451 on Build GRCh38
rs768606668 Chr.14: 104773562-104773562 on Build GRCh38
rs919281468 Chr.14: 104773645-104773645 on Build GRCh38
rs919368933 Chr.14: 104773703-104773703 on Build GRCh38
rs745809388 Chr.14: 104773537-104773537 on Build GRCh38
rs370490407 Chr.14: 104773445-104773445 on Build GRCh38
rs756044123 Chr.14: 104773617-104773617 on Build GRCh38
rs751144650 Chr.14: 104773604-104773604 on Build GRCh38
rs371467719 Chr.14: 104773497-104773497 on Build GRCh38
rs976203905 Chr.14: 104773700-104773700 on Build GRCh38
rs749939663 Chr.14: 104773697-104773697 on Build GRCh38
rs1002728299 Chr.14: 104773804-104773804 on Build GRCh38
rs754317207 Chr.14: 104773493-104773493 on Build GRCh38
rs777252096 Chr.14: 104773441-104773441 on Build GRCh38
rs377077146 Chr.14: 104773585-104773585 on Build GRCh38
rs34402148 Chr.14: 104773422-104773422 on Build GRCh38
rs773520823 Chr.14: 104773564-104773564 on Build GRCh38
rs780571834 Chr.14: 104773532-104773532 on Build GRCh38
rs897920742 Chr.14: 104773436-104773436 on Build GRCh38
rs753076466 Chr.14: 104773592-104773592 on Build GRCh38
rs749170452 Chr.14: 104773430-104773430 on Build GRCh38
rs370808079 Chr.14: 104773444-104773444 on Build GRCh38
rs558533566 Chr.14: 104773811-104773811 on Build GRCh38
rs201611116 Chr.14: 104773440-104773440 on Build GRCh38
rs564652720 Chr.14: 104773410-104773410 on Build GRCh38
rs3730328 Chr.14: 104773705-104773705 on Build GRCh38
rs909167497 Chr.14: 104773735-104773735 on Build GRCh38
rs779874420 Chr.14: 104773547-104773547 on Build GRCh38
rs968336143 Chr.14: 104773661-104773661 on Build GRCh38
rs367980124 Chr.14: 104773420-104773420 on Build GRCh38
rs756697784 Chr.14: 104773531-104773531 on Build GRCh38
rs3730362 Chr.14: 104773774-104773774 on Build GRCh38
rs779162518 Chr.14: 104773424-104773424 on Build GRCh38
rs958443676 Chr.14: 104773809-104773809 on Build GRCh38
rs747771020 Chr.14: 104773433-104773433 on Build GRCh38
rs561463952 Chr.14: 104773698-104773698 on Build GRCh38
rs566976299 Chr.14: 104773672-104773672 on Build GRCh38
rs770980034 Chr.14: 104773580-104773580 on Build GRCh38
rs55874915 Chr.14: 104773663-104773663 on Build GRCh38
rs766397090 Chr.14: 104773719-104773719 on Build GRCh38
rs779068915 Chr.14: 104773680-104773680 on Build GRCh38
rs775212616 Chr.14: 104773455-104773455 on Build GRCh38
rs571107606 Chr.14: 104773616-104773616 on Build GRCh38
rs769090254 Chr.14: 104773578-104773578 on Build GRCh38
rs549083521 Chr.14: 104773486-104773486 on Build GRCh38
rs199937067 Chr.14: 104773609-104773609 on Build GRCh38
rs375395037 Chr.14: 104773520-104773520 on Build GRCh38
rs946932902 Chr.14: 104773430-104773430 on Build GRCh38
rs866189819 Chr.14: 104773773-104773773 on Build GRCh38
rs143933508 Chr.14: 104773500-104773500 on Build GRCh38
rs930647354 Chr.14: 104773652-104773653 on Build GRCh38
rs367635775 Chr.14: 104773485-104773485 on Build GRCh38
rs762108714 Chr.14: 104773464-104773464 on Build GRCh38
rs777816547 Chr.14: 104773426-104773437 on Build GRCh38
rs368667384 Chr.14: 104773545-104773545 on Build GRCh38
rs982744857 Chr.14: 104773650-104773650 on Build GRCh38
rs759966382 Chr.14: 104773494-104773494 on Build GRCh38
rs764594578 Chr.14: 104773668-104773668 on Build GRCh38
rs907104396 Chr.14: 104773799-104773799 on Build GRCh38
rs2230508 Chr.14: 104773473-104773473 on Build GRCh38
rs758025607 Chr.14: 104773502-104773502 on Build GRCh38
rs754981294 Chr.14: 104773387-104773387 on Build GRCh38
rs1130233 Chr.14: 104773557-104773557 on Build GRCh38
rs746561686 Chr.14: 104773442-104773442 on Build GRCh38
rs1020141122 Chr.14: 104773769-104773769 on Build GRCh38
rs767037102 Chr.14: 104773409-104773409 on Build GRCh38
rs763253068 Chr.14: 104773596-104773596 on Build GRCh38
rs751232562 Chr.14: 104773525-104773525 on Build GRCh38
rs376072144 Chr.14: 104773437-104773437 on Build GRCh38
rs556745881 Chr.14: 104773635-104773635 on Build GRCh38
rs780736017 Chr.14: 104773610-104773610 on Build GRCh38
rs747036873 Chr.14: 104773383-104773383 on Build GRCh38
rs1044369768 Chr.14: 104773788-104773788 on Build GRCh38
rs756663391 Chr.14: 104773383-104773383 on Build GRCh38

View Full Product Details