Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21311867-21312365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
UniGene
Hs.514012
Gene Chromosome Location
Chr.17: 21284656-21315240 on Build GRCh38
Transcripts
NM_001316332.1 NM_002756.4 NM_145109.2 XM_005256723.2 XM_011523958.2 XM_011523959.1 XM_017024857.1 XM_017024858.1 XM_017024859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4264761 c.706C>T p.R236W Chr.17: 21312160-21312160 on Build GRCh38
6025882 c.848A>T p.E283V Chr.17: 21312215-21312215 on Build GRCh38
6025884 c.761A>T p.E254V Chr.17: 21312215-21312215 on Build GRCh38
6706953 c.874G>A p.D292N Chr.17: 21312241-21312241 on Build GRCh38
7427491 c.878G>A p.R293H Chr.17: 21312245-21312245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911373397 Chr.17: 21311896-21311896 on Build GRCh38
rs754594634 Chr.17: 21312125-21312126 on Build GRCh38
rs776797051 Chr.17: 21312165-21312165 on Build GRCh38
rs770932904 Chr.17: 21312200-21312200 on Build GRCh38
rs55736474 Chr.17: 21312225-21312225 on Build GRCh38
rs376891281 Chr.17: 21312217-21312217 on Build GRCh38
rs376993160 Chr.17: 21312222-21312222 on Build GRCh38
rs762248130 Chr.17: 21312292-21312292 on Build GRCh38
rs73982796 Chr.17: 21311932-21311932 on Build GRCh38
rs62055361 Chr.17: 21311985-21311985 on Build GRCh38
rs778493330 Chr.17: 21312147-21312147 on Build GRCh38
rs56369732 Chr.17: 21312240-21312240 on Build GRCh38
rs71371053 Chr.17: 21312157-21312157 on Build GRCh38
rs776367247 Chr.17: 21312201-21312201 on Build GRCh38
rs772054686 Chr.17: 21312198-21312200 on Build GRCh38
rs113488837 Chr.17: 21312076-21312076 on Build GRCh38
rs747865351 Chr.17: 21312189-21312189 on Build GRCh38
rs762040308 Chr.17: 21312227-21312227 on Build GRCh38
rs778314866 Chr.17: 21312127-21312128 on Build GRCh38
rs66486636 Chr.17: 21312325-21312325 on Build GRCh38
rs781026403 Chr.17: 21312252-21312252 on Build GRCh38
rs772881322 Chr.17: 21312161-21312161 on Build GRCh38
rs768070212 Chr.17: 21312232-21312232 on Build GRCh38
rs866227999 Chr.17: 21312068-21312068 on Build GRCh38
rs561081251 Chr.17: 21312246-21312246 on Build GRCh38
rs749329015 Chr.17: 21312270-21312270 on Build GRCh38
rs753143186 Chr.17: 21312124-21312124 on Build GRCh38
rs1017197686 Chr.17: 21312071-21312071 on Build GRCh38
rs538777379 Chr.17: 21312123-21312123 on Build GRCh38
rs756613703 Chr.17: 21312239-21312239 on Build GRCh38
rs9909362 Chr.17: 21311892-21311892 on Build GRCh38
rs762383350 Chr.17: 21312308-21312308 on Build GRCh38
rs907238495 Chr.17: 21312020-21312020 on Build GRCh38
rs771389358 Chr.17: 21312128-21312128 on Build GRCh38
rs151338667 Chr.17: 21312186-21312186 on Build GRCh38
rs35206134 Chr.17: 21312245-21312245 on Build GRCh38
rs73982795 Chr.17: 21311899-21311899 on Build GRCh38
rs746373047 Chr.17: 21312178-21312179 on Build GRCh38
rs67669954 Chr.17: 21312123-21312124 on Build GRCh38
rs745957434 Chr.17: 21312214-21312214 on Build GRCh38
rs368095963 Chr.17: 21312175-21312175 on Build GRCh38
rs773538684 Chr.17: 21312303-21312303 on Build GRCh38
rs113634008 Chr.17: 21312122-21312123 on Build GRCh38
rs370462268 Chr.17: 21312253-21312253 on Build GRCh38
rs753415767 Chr.17: 21312181-21312181 on Build GRCh38
rs757211923 Chr.17: 21312120-21312121 on Build GRCh38
rs1002954237 Chr.17: 21312027-21312027 on Build GRCh38
rs73302034 Chr.17: 21312331-21312331 on Build GRCh38
rs199528658 Chr.17: 21312135-21312135 on Build GRCh38
rs532738416 Chr.17: 21311944-21311944 on Build GRCh38
rs752164959 Chr.17: 21312132-21312132 on Build GRCh38
rs1055048064 Chr.17: 21311939-21311939 on Build GRCh38
rs766824035 Chr.17: 21312305-21312305 on Build GRCh38
rs747011309 Chr.17: 21312162-21312162 on Build GRCh38
rs62055360 Chr.17: 21311951-21311951 on Build GRCh38
rs771917013 Chr.17: 21312154-21312154 on Build GRCh38
rs780886953 Chr.17: 21312333-21312333 on Build GRCh38
rs117486238 Chr.17: 21312034-21312034 on Build GRCh38
rs62055363 Chr.17: 21312134-21312134 on Build GRCh38
rs775570062 Chr.17: 21312278-21312279 on Build GRCh38
rs768951364 Chr.17: 21312275-21312275 on Build GRCh38
rs775065692 Chr.17: 21312176-21312219 on Build GRCh38
rs749034663 Chr.17: 21312102-21312102 on Build GRCh38
rs758017658 Chr.17: 21312195-21312195 on Build GRCh38
rs763822224 Chr.17: 21312319-21312319 on Build GRCh38
rs758769412 Chr.17: 21312128-21312128 on Build GRCh38
rs35026048 Chr.17: 21312112-21312112 on Build GRCh38
rs770752770 Chr.17: 21312164-21312164 on Build GRCh38
rs914808575 Chr.17: 21311950-21311950 on Build GRCh38
rs375249695 Chr.17: 21312304-21312304 on Build GRCh38
rs750953487 Chr.17: 21312237-21312237 on Build GRCh38
rs548970459 Chr.17: 21312035-21312035 on Build GRCh38
rs769820739 Chr.17: 21312258-21312258 on Build GRCh38
rs116333907 Chr.17: 21311925-21311925 on Build GRCh38
rs768263661 Chr.17: 21312111-21312111 on Build GRCh38
rs780328276 Chr.17: 21312262-21312262 on Build GRCh38
rs553220653 Chr.17: 21312094-21312094 on Build GRCh38
rs572893701 Chr.17: 21312256-21312256 on Build GRCh38
rs758070523 Chr.17: 21312242-21312242 on Build GRCh38
rs778437382 Chr.17: 21312129-21312129 on Build GRCh38
rs879625738 Chr.17: 21312008-21312008 on Build GRCh38
rs1012050271 Chr.17: 21312053-21312053 on Build GRCh38
rs772387892 Chr.17: 21312296-21312296 on Build GRCh38
rs754414832 Chr.17: 21312185-21312185 on Build GRCh38
rs767860294 Chr.17: 21312179-21312179 on Build GRCh38
rs757699361 Chr.17: 21312133-21312133 on Build GRCh38
rs760520096 Chr.17: 21312121-21312121 on Build GRCh38
rs777673763 Chr.17: 21312196-21312196 on Build GRCh38
rs375515698 Chr.17: 21312136-21312136 on Build GRCh38
rs769290368 Chr.17: 21312093-21312093 on Build GRCh38
rs371545792 Chr.17: 21312221-21312221 on Build GRCh38
rs763253789 Chr.17: 21312174-21312174 on Build GRCh38
rs751837128 Chr.17: 21312176-21312176 on Build GRCh38
rs773079945 Chr.17: 21312118-21312118 on Build GRCh38
rs942913050 Chr.17: 21311898-21311898 on Build GRCh38
rs796957766 Chr.17: 21312127-21312128 on Build GRCh38
rs747953914 Chr.17: 21312153-21312153 on Build GRCh38
rs55935757 Chr.17: 21312171-21312171 on Build GRCh38
rs777252658 Chr.17: 21312244-21312244 on Build GRCh38
rs28563020 Chr.17: 21312083-21312083 on Build GRCh38
rs772025236 Chr.17: 21312117-21312117 on Build GRCh38
rs765561073 Chr.17: 21312170-21312170 on Build GRCh38
rs774365608 Chr.17: 21312283-21312283 on Build GRCh38
rs189331942 Chr.17: 21311984-21311984 on Build GRCh38

View Full Product Details