Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 21313707-21314230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP2K3
Entrez Gene ID
5606
Gene Name
mitogen-activated protein kinase kinase 3
Gene Aliases
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
UniGene
Hs.514012
Gene Chromosome Location
Chr.17: 21284656-21315240 on Build GRCh38
Transcripts
NM_001316332.1 NM_002756.4 NM_145109.2 XM_005256723.2 XM_011523958.2 XM_011523959.1 XM_017024857.1 XM_017024858.1 XM_017024859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6145890 c.991T>A p.L331M Chr.17: 21314165-21314165 on Build GRCh38
1646778 c.890C>T p.T297I Chr.17: 21314163-21314163 on Build GRCh38
6145888 c.913G>A p.D305N Chr.17: 21314186-21314186 on Build GRCh38
3515156 c.1005G>A p.K335K Chr.17: 21314179-21314179 on Build GRCh38
6145887 c.1012G>A p.D338N Chr.17: 21314186-21314186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149799475 Chr.17: 21314169-21314169 on Build GRCh38
rs766473365 Chr.17: 21314193-21314193 on Build GRCh38
rs769917287 Chr.17: 21314141-21314141 on Build GRCh38
rs767643229 Chr.17: 21314144-21314144 on Build GRCh38
rs539939314 Chr.17: 21314042-21314042 on Build GRCh38
rs4114290 Chr.17: 21313946-21313946 on Build GRCh38
rs780805421 Chr.17: 21314202-21314202 on Build GRCh38
rs190380253 Chr.17: 21313895-21313895 on Build GRCh38
rs1026555662 Chr.17: 21313758-21313758 on Build GRCh38
rs776289984 Chr.17: 21314107-21314107 on Build GRCh38
rs542399514 Chr.17: 21313752-21313752 on Build GRCh38
rs780565622 Chr.17: 21314173-21314173 on Build GRCh38
rs761720851 Chr.17: 21314189-21314189 on Build GRCh38
rs560690826 Chr.17: 21313844-21313844 on Build GRCh38
rs751670169 Chr.17: 21314140-21314140 on Build GRCh38
rs780504964 Chr.17: 21314153-21314153 on Build GRCh38
rs568772109 Chr.17: 21314116-21314116 on Build GRCh38
rs781713524 Chr.17: 21313937-21313937 on Build GRCh38
rs201112044 Chr.17: 21314179-21314179 on Build GRCh38
rs756439084 Chr.17: 21313734-21313734 on Build GRCh38
rs765812152 Chr.17: 21314152-21314152 on Build GRCh38
rs978661926 Chr.17: 21314086-21314086 on Build GRCh38
rs1018901679 Chr.17: 21313880-21313880 on Build GRCh38
rs916958249 Chr.17: 21314113-21314113 on Build GRCh38
rs1622401 Chr.17: 21314088-21314088 on Build GRCh38
rs375998287 Chr.17: 21314125-21314125 on Build GRCh38
rs767699965 Chr.17: 21314191-21314191 on Build GRCh38
rs780267199 Chr.17: 21313772-21313772 on Build GRCh38
rs753324184 Chr.17: 21314200-21314200 on Build GRCh38
rs1673837 Chr.17: 21313978-21313978 on Build GRCh38
rs181495822 Chr.17: 21314072-21314072 on Build GRCh38
rs755339514 Chr.17: 21314197-21314197 on Build GRCh38
rs761814507 Chr.17: 21314142-21314142 on Build GRCh38
rs1657684 Chr.17: 21314099-21314099 on Build GRCh38
rs749870564 Chr.17: 21314096-21314096 on Build GRCh38
rs547952800 Chr.17: 21314166-21314166 on Build GRCh38
rs774212609 Chr.17: 21314186-21314186 on Build GRCh38
rs1008827938 Chr.17: 21313846-21313846 on Build GRCh38
rs878975671 Chr.17: 21313926-21313927 on Build GRCh38
rs974887220 Chr.17: 21313924-21313924 on Build GRCh38
rs371169847 Chr.17: 21314185-21314185 on Build GRCh38
rs375674385 Chr.17: 21314073-21314073 on Build GRCh38
rs760983046 Chr.17: 21314192-21314192 on Build GRCh38
rs386796140 Chr.17: 21313926-21313927 on Build GRCh38
rs754209897 Chr.17: 21314163-21314163 on Build GRCh38
rs116715468 Chr.17: 21314164-21314164 on Build GRCh38
rs1657685 Chr.17: 21314085-21314085 on Build GRCh38
rs35755743 Chr.17: 21314184-21314184 on Build GRCh38
rs1059421 Chr.17: 21314207-21314207 on Build GRCh38
rs777451082 Chr.17: 21314100-21314100 on Build GRCh38
rs575210798 Chr.17: 21313727-21313727 on Build GRCh38
rs748542798 Chr.17: 21314013-21314013 on Build GRCh38
rs759097273 Chr.17: 21314111-21314111 on Build GRCh38
rs2363198 Chr.17: 21314201-21314201 on Build GRCh38
rs200084112 Chr.17: 21314112-21314112 on Build GRCh38
rs750711278 Chr.17: 21314152-21314152 on Build GRCh38
rs1673838 Chr.17: 21313926-21313926 on Build GRCh38
rs1044951579 Chr.17: 21313728-21313728 on Build GRCh38
rs770716588 Chr.17: 21314106-21314106 on Build GRCh38
rs186277389 Chr.17: 21314196-21314196 on Build GRCh38
rs769307504 Chr.17: 21314183-21314183 on Build GRCh38
rs965121824 Chr.17: 21313915-21313916 on Build GRCh38
rs746482925 Chr.17: 21314102-21314102 on Build GRCh38
rs886707215 Chr.17: 21313777-21313777 on Build GRCh38
rs4114291 Chr.17: 21313927-21313927 on Build GRCh38
rs75259243 Chr.17: 21313962-21313962 on Build GRCh38
rs770532577 Chr.17: 21314170-21314170 on Build GRCh38
rs765684474 Chr.17: 21314198-21314198 on Build GRCh38
rs1028571286 Chr.17: 21313944-21313944 on Build GRCh38

View Full Product Details