Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159314648-159315144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
UniGene
Hs.674
Gene Chromosome Location
Chr.5: 159314783-159330473 on Build GRCh38
Transcripts
NM_002187.2

SNP Detail

SNP ID Chromosome Location
rs774569234 Chr.5: 159314904-159314904 on Build GRCh38
rs560735709 Chr.5: 159314874-159314874 on Build GRCh38
rs56081148 Chr.5: 159315110-159315110 on Build GRCh38
rs1007051878 Chr.5: 159314755-159314755 on Build GRCh38
rs753825710 Chr.5: 159314749-159314749 on Build GRCh38
rs1004405276 Chr.5: 159314979-159314979 on Build GRCh38
rs562878121 Chr.5: 159314907-159314907 on Build GRCh38
rs527479600 Chr.5: 159315032-159315032 on Build GRCh38
rs994513840 Chr.5: 159315044-159315044 on Build GRCh38
rs916498221 Chr.5: 159314893-159314893 on Build GRCh38
rs1368439 Chr.5: 159315006-159315006 on Build GRCh38
rs1007494016 Chr.5: 159315082-159315082 on Build GRCh38
rs952716404 Chr.5: 159314977-159314977 on Build GRCh38
rs1029877458 Chr.5: 159314714-159314714 on Build GRCh38
rs890031640 Chr.5: 159315045-159315045 on Build GRCh38
rs540857937 Chr.5: 159314859-159314859 on Build GRCh38
rs531770868 Chr.5: 159314909-159314909 on Build GRCh38
rs547204212 Chr.5: 159315085-159315085 on Build GRCh38
rs951662271 Chr.5: 159314766-159314766 on Build GRCh38
rs944241542 Chr.5: 159315019-159315019 on Build GRCh38
rs369997793 Chr.5: 159314962-159314962 on Build GRCh38
rs963040390 Chr.5: 159314826-159314829 on Build GRCh38
rs886060353 Chr.5: 159315019-159315019 on Build GRCh38
rs962979200 Chr.5: 159315024-159315024 on Build GRCh38
rs998717007 Chr.5: 159314957-159314957 on Build GRCh38
rs926061045 Chr.5: 159314931-159314931 on Build GRCh38
rs1053101720 Chr.5: 159314953-159314953 on Build GRCh38
rs927070878 Chr.5: 159314837-159314837 on Build GRCh38
rs918269319 Chr.5: 159314833-159314833 on Build GRCh38
rs976881088 Chr.5: 159315022-159315022 on Build GRCh38
rs575691756 Chr.5: 159314734-159314734 on Build GRCh38
rs886060351 Chr.5: 159314826-159314829 on Build GRCh38
rs936103986 Chr.5: 159314948-159314948 on Build GRCh38
rs555485221 Chr.5: 159314765-159314765 on Build GRCh38
rs958742642 Chr.5: 159314842-159314845 on Build GRCh38
rs868312621 Chr.5: 159314798-159314798 on Build GRCh38
rs766284185 Chr.5: 159314683-159314683 on Build GRCh38
rs559187957 Chr.5: 159314921-159314921 on Build GRCh38
rs1040160247 Chr.5: 159314946-159314946 on Build GRCh38
rs1017133453 Chr.5: 159315095-159315095 on Build GRCh38
rs886060352 Chr.5: 159314869-159314869 on Build GRCh38
rs181807125 Chr.5: 159314915-159314915 on Build GRCh38
rs954397822 Chr.5: 159314729-159314729 on Build GRCh38
rs574914587 Chr.5: 159314789-159314789 on Build GRCh38
rs992670509 Chr.5: 159314861-159314861 on Build GRCh38
rs1039808375 Chr.5: 159314731-159314731 on Build GRCh38
rs149441492 Chr.5: 159314758-159314758 on Build GRCh38
rs546174255 Chr.5: 159314901-159314901 on Build GRCh38
rs144817848 Chr.5: 159314879-159314879 on Build GRCh38
rs1014531642 Chr.5: 159314825-159314826 on Build GRCh38
rs1051266390 Chr.5: 159314971-159314971 on Build GRCh38
rs1000261829 Chr.5: 159314712-159314712 on Build GRCh38
rs998372927 Chr.5: 159314747-159314747 on Build GRCh38
rs998196106 Chr.5: 159315005-159315005 on Build GRCh38
rs908778623 Chr.5: 159315013-159315013 on Build GRCh38
rs754804497 Chr.5: 159314986-159314986 on Build GRCh38

View Full Product Details