Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159318698-159319246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
UniGene
Hs.674
Gene Chromosome Location
Chr.5: 159314783-159330473 on Build GRCh38
Transcripts
NM_002187.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6867420 c.813G>A p.L271L Chr.5: 159318778-159318778 on Build GRCh38
3613382 c.810C>T p.S270S Chr.5: 159318781-159318781 on Build GRCh38
5402916 c.794C>T p.P265L Chr.5: 159318797-159318797 on Build GRCh38
4585670 c.749G>A p.R250Q Chr.5: 159318842-159318842 on Build GRCh38
7143253 c.840G>A p.K280K Chr.5: 159318751-159318751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200529978 Chr.5: 159318907-159318907 on Build GRCh38
rs773370181 Chr.5: 159318919-159318919 on Build GRCh38
rs778843948 Chr.5: 159318861-159318861 on Build GRCh38
rs575161274 Chr.5: 159318869-159318869 on Build GRCh38
rs770877646 Chr.5: 159318751-159318751 on Build GRCh38
rs766902711 Chr.5: 159318829-159318829 on Build GRCh38
rs745818512 Chr.5: 159318872-159318872 on Build GRCh38
rs192540461 Chr.5: 159318853-159318853 on Build GRCh38
rs768677098 Chr.5: 159318883-159318883 on Build GRCh38
rs372475701 Chr.5: 159318762-159318762 on Build GRCh38
rs376252390 Chr.5: 159318908-159318908 on Build GRCh38
rs761924828 Chr.5: 159318903-159318903 on Build GRCh38
rs372926766 Chr.5: 159318855-159318855 on Build GRCh38
rs922166421 Chr.5: 159319096-159319096 on Build GRCh38
rs776775490 Chr.5: 159318775-159318775 on Build GRCh38
rs551633381 Chr.5: 159319077-159319077 on Build GRCh38
rs989927547 Chr.5: 159319002-159319002 on Build GRCh38
rs750772366 Chr.5: 159318821-159318821 on Build GRCh38
rs189324104 Chr.5: 159318768-159318768 on Build GRCh38
rs111492985 Chr.5: 159318740-159318740 on Build GRCh38
rs919394608 Chr.5: 159319188-159319188 on Build GRCh38
rs1049789144 Chr.5: 159318887-159318887 on Build GRCh38
rs543882576 Chr.5: 159318949-159318949 on Build GRCh38
rs775647036 Chr.5: 159319076-159319076 on Build GRCh38
rs1049275961 Chr.5: 159319189-159319189 on Build GRCh38
rs1009458305 Chr.5: 159319039-159319039 on Build GRCh38
rs946817426 Chr.5: 159319042-159319042 on Build GRCh38
rs932162610 Chr.5: 159319177-159319177 on Build GRCh38
rs763308156 Chr.5: 159318921-159318921 on Build GRCh38
rs758172137 Chr.5: 159319222-159319222 on Build GRCh38
rs765482576 Chr.5: 159318813-159318813 on Build GRCh38
rs904934986 Chr.5: 159318996-159318996 on Build GRCh38
rs368420106 Chr.5: 159319176-159319176 on Build GRCh38
rs753138960 Chr.5: 159318929-159318929 on Build GRCh38
rs762049572 Chr.5: 159318782-159318782 on Build GRCh38
rs752581359 Chr.5: 159319056-159319056 on Build GRCh38
rs898029732 Chr.5: 159318881-159318881 on Build GRCh38
rs528709369 Chr.5: 159319065-159319065 on Build GRCh38
rs368458310 Chr.5: 159318843-159318843 on Build GRCh38
rs776726308 Chr.5: 159318888-159318888 on Build GRCh38
rs559514707 Chr.5: 159319053-159319053 on Build GRCh38
rs754475899 Chr.5: 159318832-159318832 on Build GRCh38
rs773089918 Chr.5: 159318729-159318729 on Build GRCh38
rs377084191 Chr.5: 159318995-159318995 on Build GRCh38
rs780867719 Chr.5: 159318833-159318833 on Build GRCh38
rs201757381 Chr.5: 159318836-159318836 on Build GRCh38
rs10036122 Chr.5: 159319206-159319206 on Build GRCh38
rs199527043 Chr.5: 159318880-159318880 on Build GRCh38
rs986537294 Chr.5: 159319119-159319119 on Build GRCh38
rs749076435 Chr.5: 159318737-159318737 on Build GRCh38
rs910589125 Chr.5: 159319139-159319139 on Build GRCh38
rs74644143 Chr.5: 159318842-159318842 on Build GRCh38
rs752059689 Chr.5: 159318933-159318933 on Build GRCh38
rs542774360 Chr.5: 159319046-159319046 on Build GRCh38
rs561010709 Chr.5: 159319000-159319000 on Build GRCh38
rs766662954 Chr.5: 159318931-159318931 on Build GRCh38
rs372906845 Chr.5: 159319209-159319209 on Build GRCh38
rs1044774952 Chr.5: 159318951-159318951 on Build GRCh38
rs752010709 Chr.5: 159318830-159318830 on Build GRCh38
rs765340406 Chr.5: 159318736-159318737 on Build GRCh38
rs764163895 Chr.5: 159318769-159318769 on Build GRCh38
rs749098724 Chr.5: 159318850-159318850 on Build GRCh38
rs1034224586 Chr.5: 159318974-159318974 on Build GRCh38

View Full Product Details