Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180240656-180241175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK9
Entrez Gene ID
5601
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK
UniGene
Hs.484371
Gene Chromosome Location
Chr.5: 180233594-180292071 on Build GRCh38
Transcripts
NM_002752.4 NM_139068.2 NM_139069.2 NM_139070.2 XM_005265940.4 XM_006714891.2 XM_017009638.1 XM_017009639.1 XM_017009640.1 XM_017009641.1 XM_017009642.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6708038 c.981C>T p.P327P Chr.5: 180241046-180241046 on Build GRCh38
3012175 c.985G>A p.E329K Chr.5: 180241042-180241042 on Build GRCh38
5767748 c.872-5C>A p.? Chr.5: 180241160-180241160 on Build GRCh38
6360596 c.898A>G p.K300E Chr.5: 180241129-180241129 on Build GRCh38
3854312 c.939C>T p.D313D Chr.5: 180241088-180241088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529831101 Chr.5: 180240771-180240771 on Build GRCh38
rs376717703 Chr.5: 180241077-180241077 on Build GRCh38
rs1044293037 Chr.5: 180240797-180240797 on Build GRCh38
rs772689390 Chr.5: 180241038-180241038 on Build GRCh38
rs767706697 Chr.5: 180241047-180241047 on Build GRCh38
rs761154333 Chr.5: 180241060-180241060 on Build GRCh38
rs79617115 Chr.5: 180241046-180241046 on Build GRCh38
rs747684606 Chr.5: 180241142-180241142 on Build GRCh38
rs992813520 Chr.5: 180241087-180241087 on Build GRCh38
rs529721305 Chr.5: 180241043-180241043 on Build GRCh38
rs112360221 Chr.5: 180240820-180240820 on Build GRCh38
rs765531121 Chr.5: 180241093-180241093 on Build GRCh38
rs754934802 Chr.5: 180241139-180241139 on Build GRCh38
rs955746941 Chr.5: 180240784-180240784 on Build GRCh38
rs764758451 Chr.5: 180240698-180240698 on Build GRCh38
rs918690590 Chr.5: 180240862-180240862 on Build GRCh38
rs747844190 Chr.5: 180241028-180241028 on Build GRCh38
rs927324582 Chr.5: 180240821-180240821 on Build GRCh38
rs938398408 Chr.5: 180240747-180240748 on Build GRCh38
rs1027250842 Chr.5: 180240918-180240918 on Build GRCh38
rs552504246 Chr.5: 180240834-180240834 on Build GRCh38
rs574819083 Chr.5: 180240912-180240912 on Build GRCh38
rs756611366 Chr.5: 180241008-180241008 on Build GRCh38
rs749684135 Chr.5: 180241138-180241138 on Build GRCh38
rs562225301 Chr.5: 180240996-180240996 on Build GRCh38
rs576369914 Chr.5: 180240995-180240995 on Build GRCh38
rs775988860 Chr.5: 180241049-180241049 on Build GRCh38
rs543847131 Chr.5: 180241003-180241003 on Build GRCh38
rs983132353 Chr.5: 180240940-180240940 on Build GRCh38
rs762603663 Chr.5: 180240747-180240747 on Build GRCh38
rs900384614 Chr.5: 180240924-180240924 on Build GRCh38
rs35621262 Chr.5: 180241042-180241043 on Build GRCh38
rs751846029 Chr.5: 180240819-180240819 on Build GRCh38
rs6895740 Chr.5: 180240933-180240933 on Build GRCh38
rs780562313 Chr.5: 180241078-180241078 on Build GRCh38
rs992752726 Chr.5: 180240855-180240855 on Build GRCh38
rs142611107 Chr.5: 180240708-180240708 on Build GRCh38
rs930044568 Chr.5: 180240872-180240872 on Build GRCh38
rs534320271 Chr.5: 180240916-180240916 on Build GRCh38
rs562837507 Chr.5: 180240975-180240975 on Build GRCh38
rs1034613831 Chr.5: 180241057-180241057 on Build GRCh38
rs776276982 Chr.5: 180241004-180241004 on Build GRCh38
rs560809759 Chr.5: 180240767-180240767 on Build GRCh38
rs569224644 Chr.5: 180240852-180240852 on Build GRCh38
rs779705113 Chr.5: 180241111-180241111 on Build GRCh38
rs530427615 Chr.5: 180240716-180240716 on Build GRCh38
rs538236012 Chr.5: 180240875-180240875 on Build GRCh38
rs758231386 Chr.5: 180241108-180241108 on Build GRCh38
rs550597412 Chr.5: 180240733-180240733 on Build GRCh38
rs546039154 Chr.5: 180240974-180240974 on Build GRCh38
rs560961564 Chr.5: 180241031-180241031 on Build GRCh38
rs769466526 Chr.5: 180241030-180241030 on Build GRCh38
rs887715751 Chr.5: 180240707-180240707 on Build GRCh38
rs750547153 Chr.5: 180241105-180241105 on Build GRCh38
rs1055577329 Chr.5: 180240752-180240752 on Build GRCh38
rs144771330 Chr.5: 180240773-180240773 on Build GRCh38
rs988996273 Chr.5: 180240798-180240798 on Build GRCh38
rs753882278 Chr.5: 180241079-180241079 on Build GRCh38
rs979601815 Chr.5: 180241118-180241118 on Build GRCh38
rs969403924 Chr.5: 180240818-180240818 on Build GRCh38
rs138900129 Chr.5: 180240884-180240884 on Build GRCh38
rs113577474 Chr.5: 180240994-180240994 on Build GRCh38
rs766297640 Chr.5: 180240694-180240694 on Build GRCh38
rs116346965 Chr.5: 180241088-180241088 on Build GRCh38
rs959733325 Chr.5: 180240961-180240961 on Build GRCh38
rs745700815 Chr.5: 180240988-180240988 on Build GRCh38
rs6894217 Chr.5: 180240806-180240806 on Build GRCh38
rs780900586 Chr.5: 180241140-180241140 on Build GRCh38
rs577992261 Chr.5: 180240874-180240874 on Build GRCh38
rs1017722809 Chr.5: 180240728-180240728 on Build GRCh38
rs201806665 Chr.5: 180241012-180241012 on Build GRCh38
rs189935590 Chr.5: 180240950-180240950 on Build GRCh38
rs951102336 Chr.5: 180241050-180241050 on Build GRCh38

View Full Product Details