Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180602269-180602773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009268.1

SNP Detail

SNP ID Chromosome Location
rs557074258 Chr.5: 180602449-180602449 on Build GRCh38
rs1031561507 Chr.5: 180602496-180602496 on Build GRCh38
rs549141464 Chr.5: 180602367-180602367 on Build GRCh38
rs911618217 Chr.5: 180602377-180602377 on Build GRCh38
rs984854763 Chr.5: 180602692-180602692 on Build GRCh38
rs969980070 Chr.5: 180602402-180602402 on Build GRCh38
rs539290222 Chr.5: 180602614-180602614 on Build GRCh38
rs74577503 Chr.5: 180602641-180602641 on Build GRCh38
rs559245198 Chr.5: 180602384-180602384 on Build GRCh38
rs1022832671 Chr.5: 180602399-180602399 on Build GRCh38
rs78447148 Chr.5: 180602670-180602670 on Build GRCh38
rs551371042 Chr.5: 180602422-180602422 on Build GRCh38
rs563268325 Chr.5: 180602690-180602690 on Build GRCh38
rs897106196 Chr.5: 180602639-180602639 on Build GRCh38
rs892878839 Chr.5: 180602537-180602537 on Build GRCh38
rs377670742 Chr.5: 180602725-180602725 on Build GRCh38
rs781498936 Chr.5: 180602300-180602300 on Build GRCh38
rs955822620 Chr.5: 180602347-180602347 on Build GRCh38
rs573585132 Chr.5: 180602606-180602606 on Build GRCh38
rs553567480 Chr.5: 180602604-180602604 on Build GRCh38
rs6877011 Chr.5: 180602471-180602471 on Build GRCh38
rs1000204443 Chr.5: 180602479-180602479 on Build GRCh38
rs940306090 Chr.5: 180602678-180602678 on Build GRCh38
rs72816989 Chr.5: 180602616-180602616 on Build GRCh38
rs757173621 Chr.5: 180602700-180602700 on Build GRCh38
rs931556469 Chr.5: 180602428-180602428 on Build GRCh38
rs987313097 Chr.5: 180602355-180602355 on Build GRCh38
rs976750123 Chr.5: 180602519-180602519 on Build GRCh38
rs947142796 Chr.5: 180602550-180602550 on Build GRCh38
rs748445442 Chr.5: 180602723-180602723 on Build GRCh38
rs537223604 Chr.5: 180602448-180602448 on Build GRCh38
rs528336613 Chr.5: 180602398-180602398 on Build GRCh38
rs922924079 Chr.5: 180602526-180602526 on Build GRCh38
rs77376470 Chr.5: 180602319-180602319 on Build GRCh38
rs536466548 Chr.5: 180602543-180602543 on Build GRCh38
rs905702457 Chr.5: 180602571-180602571 on Build GRCh38
rs761273261 Chr.5: 180602665-180602665 on Build GRCh38
rs1052857627 Chr.5: 180602343-180602343 on Build GRCh38
rs960996460 Chr.5: 180602439-180602439 on Build GRCh38
rs1013035363 Chr.5: 180602459-180602459 on Build GRCh38
rs145794559 Chr.5: 180602656-180602659 on Build GRCh38
rs955956621 Chr.5: 180602513-180602513 on Build GRCh38
rs934929278 Chr.5: 180602750-180602750 on Build GRCh38
rs969030416 Chr.5: 180602301-180602301 on Build GRCh38
rs1052803057 Chr.5: 180602534-180602534 on Build GRCh38
rs1026350699 Chr.5: 180602441-180602441 on Build GRCh38
rs189061036 Chr.5: 180602440-180602440 on Build GRCh38
rs371593506 Chr.5: 180602658-180602661 on Build GRCh38
rs76601685 Chr.5: 180602746-180602746 on Build GRCh38
rs139719678 Chr.5: 180602724-180602724 on Build GRCh38
rs77831530 Chr.5: 180602458-180602458 on Build GRCh38
rs1017739526 Chr.5: 180602722-180602722 on Build GRCh38
rs1003218651 Chr.5: 180602407-180602407 on Build GRCh38
rs934317630 Chr.5: 180602529-180602529 on Build GRCh38
rs1002758085 Chr.5: 180602575-180602575 on Build GRCh38
rs998582061 Chr.5: 180602324-180602324 on Build GRCh38
rs550828520 Chr.5: 180602460-180602460 on Build GRCh38
rs992949234 Chr.5: 180602682-180602682 on Build GRCh38
rs1044225452 Chr.5: 180602564-180602564 on Build GRCh38

View Full Product Details