Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154429118-154429614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL6R
Entrez Gene ID
3570
Gene Name
interleukin 6 receptor
Gene Aliases
CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
UniGene
Hs.135087
Gene Chromosome Location
Chr.1: 154405193-154469450 on Build GRCh38
Transcripts
NM_000565.3 NM_001206866.1 NM_181359.2 XM_005245139.1 XM_005245140.2 XM_006711298.1 XM_006711299.3 XM_017001199.1 XM_017001200.1 XM_017001201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5759846 c.112C>T p.L38L Chr.1: 154429222-154429222 on Build GRCh38
4622131 c.203G>A p.G68D Chr.1: 154429313-154429313 on Build GRCh38
5727852 c.294G>A p.R98R Chr.1: 154429404-154429404 on Build GRCh38
4613257 c.144delG p.V50fs*1 Chr.1: 154429254-154429254 on Build GRCh38
3474681 c.188G>A p.R63K Chr.1: 154429298-154429298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750083210 Chr.1: 154429257-154429257 on Build GRCh38
rs113787847 Chr.1: 154429395-154429395 on Build GRCh38
rs752579637 Chr.1: 154429222-154429222 on Build GRCh38
rs1044860196 Chr.1: 154429196-154429196 on Build GRCh38
rs981512355 Chr.1: 154429524-154429524 on Build GRCh38
rs969444010 Chr.1: 154429500-154429500 on Build GRCh38
rs759668639 Chr.1: 154429407-154429407 on Build GRCh38
rs377635182 Chr.1: 154429475-154429475 on Build GRCh38
rs773375947 Chr.1: 154429315-154429315 on Build GRCh38
rs145156061 Chr.1: 154429408-154429408 on Build GRCh38
rs994295810 Chr.1: 154429170-154429170 on Build GRCh38
rs868345266 Chr.1: 154429202-154429202 on Build GRCh38
rs748064464 Chr.1: 154429480-154429480 on Build GRCh38
rs767071743 Chr.1: 154429429-154429429 on Build GRCh38
rs376202798 Chr.1: 154429145-154429145 on Build GRCh38
rs769250668 Chr.1: 154429295-154429295 on Build GRCh38
rs34099703 Chr.1: 154429236-154429236 on Build GRCh38
rs774440162 Chr.1: 154429333-154429333 on Build GRCh38
rs942306855 Chr.1: 154429552-154429552 on Build GRCh38
rs539803011 Chr.1: 154429467-154429467 on Build GRCh38
rs779555183 Chr.1: 154429274-154429274 on Build GRCh38
rs775930595 Chr.1: 154429190-154429190 on Build GRCh38
rs576477390 Chr.1: 154429502-154429502 on Build GRCh38
rs147437437 Chr.1: 154429310-154429310 on Build GRCh38
rs771599478 Chr.1: 154429189-154429189 on Build GRCh38
rs919202342 Chr.1: 154429593-154429593 on Build GRCh38
rs143539244 Chr.1: 154429566-154429566 on Build GRCh38
rs1036259781 Chr.1: 154429378-154429378 on Build GRCh38
rs373941142 Chr.1: 154429412-154429412 on Build GRCh38
rs756596344 Chr.1: 154429435-154429435 on Build GRCh38
rs766108946 Chr.1: 154429417-154429417 on Build GRCh38
rs980955125 Chr.1: 154429510-154429510 on Build GRCh38
rs774533163 Chr.1: 154429405-154429405 on Build GRCh38
rs146168222 Chr.1: 154429281-154429281 on Build GRCh38
rs745420665 Chr.1: 154429164-154429164 on Build GRCh38
rs11557725 Chr.1: 154429352-154429352 on Build GRCh38
rs758016763 Chr.1: 154429371-154429371 on Build GRCh38
rs757025871 Chr.1: 154429237-154429237 on Build GRCh38
rs2228144 Chr.1: 154429203-154429203 on Build GRCh38
rs147678204 Chr.1: 154429349-154429349 on Build GRCh38
rs6694817 Chr.1: 154429496-154429496 on Build GRCh38
rs765013919 Chr.1: 154429364-154429364 on Build GRCh38
rs768837795 Chr.1: 154429564-154429564 on Build GRCh38
rs545906094 Chr.1: 154429459-154429459 on Build GRCh38
rs369317343 Chr.1: 154429380-154429380 on Build GRCh38
rs778489682 Chr.1: 154429242-154429242 on Build GRCh38
rs748953191 Chr.1: 154429148-154429148 on Build GRCh38
rs773005536 Chr.1: 154429486-154429486 on Build GRCh38
rs766474176 Chr.1: 154429216-154429216 on Build GRCh38
rs778598316 Chr.1: 154429161-154429161 on Build GRCh38
rs747537633 Chr.1: 154429278-154429278 on Build GRCh38
rs761749675 Chr.1: 154429411-154429411 on Build GRCh38
rs749709843 Chr.1: 154429456-154429456 on Build GRCh38
rs749494588 Chr.1: 154429403-154429403 on Build GRCh38
rs138458259 Chr.1: 154429265-154429265 on Build GRCh38
rs748663885 Chr.1: 154429390-154429390 on Build GRCh38
rs764067079 Chr.1: 154429340-154429340 on Build GRCh38
rs368041955 Chr.1: 154429481-154429481 on Build GRCh38
rs200004280 Chr.1: 154429410-154429410 on Build GRCh38
rs929657462 Chr.1: 154429583-154429583 on Build GRCh38
rs756864381 Chr.1: 154429153-154429153 on Build GRCh38
rs367578267 Chr.1: 154429307-154429307 on Build GRCh38
rs370155930 Chr.1: 154429402-154429402 on Build GRCh38
rs775496278 Chr.1: 154429478-154429478 on Build GRCh38
rs770930455 Chr.1: 154429328-154429328 on Build GRCh38
rs1016993782 Chr.1: 154429144-154429144 on Build GRCh38
rs759580674 Chr.1: 154429334-154429334 on Build GRCh38
rs538533109 Chr.1: 154429209-154429209 on Build GRCh38
rs572856067 Chr.1: 154429428-154429428 on Build GRCh38
rs748575982 Chr.1: 154429305-154429305 on Build GRCh38
rs750053703 Chr.1: 154429365-154429365 on Build GRCh38
rs78133617 Chr.1: 154429304-154429304 on Build GRCh38
rs747456382 Chr.1: 154429195-154429195 on Build GRCh38
rs762997891 Chr.1: 154429215-154429215 on Build GRCh38
rs762628881 Chr.1: 154429416-154429416 on Build GRCh38
rs909352035 Chr.1: 154429525-154429525 on Build GRCh38
rs777476553 Chr.1: 154429146-154429146 on Build GRCh38
rs748799521 Chr.1: 154429148-154429149 on Build GRCh38
rs751246387 Chr.1: 154429425-154429425 on Build GRCh38
rs145766309 Chr.1: 154429382-154429382 on Build GRCh38
rs753271783 Chr.1: 154429430-154429430 on Build GRCh38
rs763907785 Chr.1: 154429235-154429235 on Build GRCh38
rs1022913861 Chr.1: 154429514-154429514 on Build GRCh38
rs770415570 Chr.1: 154429254-154429254 on Build GRCh38
rs765593427 Chr.1: 154429212-154429212 on Build GRCh38
rs1019106367 Chr.1: 154429296-154429296 on Build GRCh38
rs773458420 Chr.1: 154429214-154429214 on Build GRCh38
rs778029868 Chr.1: 154429454-154429454 on Build GRCh38
rs774333916 Chr.1: 154429253-154429254 on Build GRCh38
rs779172175 Chr.1: 154429465-154429465 on Build GRCh38
rs199686914 Chr.1: 154429351-154429351 on Build GRCh38
rs2229237 Chr.1: 154429320-154429320 on Build GRCh38
rs779457451 Chr.1: 154429376-154429376 on Build GRCh38
rs544082222 Chr.1: 154429520-154429520 on Build GRCh38
rs994608378 Chr.1: 154429219-154429219 on Build GRCh38
rs142365714 Chr.1: 154429377-154429377 on Build GRCh38
rs376818209 Chr.1: 154429466-154429466 on Build GRCh38
rs915091270 Chr.1: 154429144-154429144 on Build GRCh38
rs762153528 Chr.1: 154429210-154429210 on Build GRCh38
rs896922982 Chr.1: 154429168-154429168 on Build GRCh38

View Full Product Details