Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63657658-63658154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K3
Entrez Gene ID
4215
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
MAPKKK3, MEKK3
UniGene
Hs.29282
Gene Chromosome Location
Chr.17: 63622209-63696310 on Build GRCh38
Transcripts
NM_002401.3 NM_203351.1 XM_005257376.3 XM_005257377.3 XM_005257378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3890155 c.365C>T p.S122F Chr.17: 63657891-63657891 on Build GRCh38
6147642 c.373C>G p.L125V Chr.17: 63657806-63657806 on Build GRCh38
6848878 c.353T>C p.I118T Chr.17: 63657879-63657879 on Build GRCh38
4700330 c.374G>T p.R125I Chr.17: 63657900-63657900 on Build GRCh38
5462680 c.370G>A p.D124N Chr.17: 63657896-63657896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917671021 Chr.17: 63657994-63657994 on Build GRCh38
rs760651201 Chr.17: 63657778-63657778 on Build GRCh38
rs935494182 Chr.17: 63657713-63657717 on Build GRCh38
rs778923849 Chr.17: 63657890-63657891 on Build GRCh38
rs908853137 Chr.17: 63658038-63658038 on Build GRCh38
rs950430390 Chr.17: 63657995-63657995 on Build GRCh38
rs749446678 Chr.17: 63658035-63658035 on Build GRCh38
rs969013008 Chr.17: 63657952-63657952 on Build GRCh38
rs760879872 Chr.17: 63657924-63657924 on Build GRCh38
rs900319073 Chr.17: 63657762-63657762 on Build GRCh38
rs894053220 Chr.17: 63657817-63657817 on Build GRCh38
rs868215387 Chr.17: 63658064-63658064 on Build GRCh38
rs757293519 Chr.17: 63657777-63657778 on Build GRCh38
rs760060741 Chr.17: 63657880-63657880 on Build GRCh38
rs16947025 Chr.17: 63657973-63657973 on Build GRCh38
rs780588217 Chr.17: 63657822-63657822 on Build GRCh38
rs941986379 Chr.17: 63658075-63658075 on Build GRCh38
rs769154360 Chr.17: 63657837-63657837 on Build GRCh38
rs983097456 Chr.17: 63658034-63658034 on Build GRCh38
rs772686374 Chr.17: 63657881-63657881 on Build GRCh38
rs975064849 Chr.17: 63658081-63658081 on Build GRCh38
rs530249887 Chr.17: 63657754-63657754 on Build GRCh38
rs933157972 Chr.17: 63658095-63658095 on Build GRCh38
rs538405229 Chr.17: 63658017-63658017 on Build GRCh38
rs921823161 Chr.17: 63658082-63658082 on Build GRCh38
rs1002182910 Chr.17: 63657956-63657956 on Build GRCh38
rs977598358 Chr.17: 63658105-63658105 on Build GRCh38
rs751348276 Chr.17: 63657957-63657957 on Build GRCh38
rs6504173 Chr.17: 63657696-63657696 on Build GRCh38
rs777548849 Chr.17: 63657840-63657840 on Build GRCh38
rs113613958 Chr.17: 63658060-63658060 on Build GRCh38
rs370404986 Chr.17: 63657761-63657761 on Build GRCh38
rs377741494 Chr.17: 63657927-63657927 on Build GRCh38
rs754924253 Chr.17: 63657810-63657810 on Build GRCh38
rs758322494 Chr.17: 63657789-63657789 on Build GRCh38
rs1034548889 Chr.17: 63657963-63657963 on Build GRCh38
rs1051842818 Chr.17: 63657841-63657841 on Build GRCh38
rs754006982 Chr.17: 63657787-63657787 on Build GRCh38
rs374735208 Chr.17: 63657872-63657872 on Build GRCh38
rs764072557 Chr.17: 63657925-63657925 on Build GRCh38
rs752776142 Chr.17: 63657776-63657776 on Build GRCh38
rs747742093 Chr.17: 63657834-63657834 on Build GRCh38
rs775832508 Chr.17: 63657907-63657907 on Build GRCh38
rs140890307 Chr.17: 63658114-63658114 on Build GRCh38
rs1010244661 Chr.17: 63657923-63657923 on Build GRCh38
rs546340382 Chr.17: 63657951-63657951 on Build GRCh38
rs148711373 Chr.17: 63657864-63657864 on Build GRCh38
rs756420339 Chr.17: 63657965-63657965 on Build GRCh38
rs767526575 Chr.17: 63657765-63657765 on Build GRCh38
rs144906918 Chr.17: 63657948-63657948 on Build GRCh38
rs748881884 Chr.17: 63657849-63657849 on Build GRCh38
rs771273140 Chr.17: 63657745-63657745 on Build GRCh38
rs751544538 Chr.17: 63657804-63657804 on Build GRCh38

View Full Product Details