Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63666862-63667374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K3
Entrez Gene ID
4215
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
MAPKKK3, MEKK3
UniGene
Hs.29282
Gene Chromosome Location
Chr.17: 63622209-63696310 on Build GRCh38
Transcripts
NM_002401.3 NM_203351.1 XM_005257376.3 XM_005257377.3 XM_005257378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3692012 c.441C>T p.S147S Chr.17: 63666999-63666999 on Build GRCh38
5573944 c.468_469insC p.E159fs*26 Chr.17: 63667026-63667027 on Build GRCh38
4269809 c.582G>T p.R194S Chr.17: 63667047-63667047 on Build GRCh38
227624 c.389C>T p.S130F Chr.17: 63666947-63666947 on Build GRCh38
3520774 c.480T>C p.S160S Chr.17: 63666945-63666945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762185565 Chr.17: 63667126-63667126 on Build GRCh38
rs778517747 Chr.17: 63666959-63666959 on Build GRCh38
rs768781864 Chr.17: 63666982-63666982 on Build GRCh38
rs34224696 Chr.17: 63667307-63667308 on Build GRCh38
rs568655432 Chr.17: 63667084-63667084 on Build GRCh38
rs904334524 Chr.17: 63667123-63667123 on Build GRCh38
rs776126405 Chr.17: 63666976-63666976 on Build GRCh38
rs752528601 Chr.17: 63666916-63666916 on Build GRCh38
rs35028835 Chr.17: 63666989-63666989 on Build GRCh38
rs756731667 Chr.17: 63667029-63667029 on Build GRCh38
rs928510869 Chr.17: 63667052-63667052 on Build GRCh38
rs767376926 Chr.17: 63667088-63667088 on Build GRCh38
rs556240867 Chr.17: 63667215-63667215 on Build GRCh38
rs572225541 Chr.17: 63667332-63667332 on Build GRCh38
rs200260705 Chr.17: 63667032-63667032 on Build GRCh38
rs775100223 Chr.17: 63667106-63667106 on Build GRCh38
rs202050738 Chr.17: 63666999-63666999 on Build GRCh38
rs767808270 Chr.17: 63667089-63667089 on Build GRCh38
rs762113008 Chr.17: 63667041-63667041 on Build GRCh38
rs568640776 Chr.17: 63666980-63666980 on Build GRCh38
rs944002195 Chr.17: 63667272-63667272 on Build GRCh38
rs576866693 Chr.17: 63667145-63667145 on Build GRCh38
rs773625164 Chr.17: 63667083-63667083 on Build GRCh38
rs752685930 Chr.17: 63667027-63667027 on Build GRCh38
rs1042910100 Chr.17: 63667113-63667116 on Build GRCh38
rs552196191 Chr.17: 63666908-63666908 on Build GRCh38
rs367914417 Chr.17: 63667050-63667050 on Build GRCh38
rs561694980 Chr.17: 63667288-63667288 on Build GRCh38
rs935515064 Chr.17: 63667306-63667306 on Build GRCh38
rs762648029 Chr.17: 63667334-63667334 on Build GRCh38
rs780212319 Chr.17: 63667038-63667038 on Build GRCh38
rs755708063 Chr.17: 63666950-63666950 on Build GRCh38
rs746273875 Chr.17: 63666969-63666969 on Build GRCh38
rs1008462811 Chr.17: 63666931-63666931 on Build GRCh38
rs539430849 Chr.17: 63667165-63667165 on Build GRCh38
rs1022609171 Chr.17: 63667031-63667031 on Build GRCh38
rs984264283 Chr.17: 63667321-63667321 on Build GRCh38
rs1041301680 Chr.17: 63667275-63667275 on Build GRCh38
rs754796483 Chr.17: 63666903-63666903 on Build GRCh38
rs751148786 Chr.17: 63667348-63667348 on Build GRCh38
rs576030933 Chr.17: 63667223-63667223 on Build GRCh38
rs749910024 Chr.17: 63667259-63667259 on Build GRCh38
rs772322521 Chr.17: 63666972-63666972 on Build GRCh38
rs1031401808 Chr.17: 63666981-63666981 on Build GRCh38
rs1051733710 Chr.17: 63667023-63667023 on Build GRCh38
rs951176144 Chr.17: 63667320-63667320 on Build GRCh38
rs920023862 Chr.17: 63667144-63667144 on Build GRCh38
rs745509910 Chr.17: 63667030-63667030 on Build GRCh38
rs376043954 Chr.17: 63666901-63666901 on Build GRCh38
rs141438505 Chr.17: 63666955-63666955 on Build GRCh38
rs911223380 Chr.17: 63667194-63667194 on Build GRCh38
rs368678801 Chr.17: 63666920-63666920 on Build GRCh38
rs115609820 Chr.17: 63667346-63667346 on Build GRCh38
rs371965403 Chr.17: 63666954-63666954 on Build GRCh38
rs534199832 Chr.17: 63667113-63667113 on Build GRCh38
rs961240075 Chr.17: 63667118-63667118 on Build GRCh38
rs1025239812 Chr.17: 63667319-63667319 on Build GRCh38
rs763617586 Chr.17: 63667028-63667028 on Build GRCh38
rs748275986 Chr.17: 63667055-63667055 on Build GRCh38
rs530203202 Chr.17: 63667286-63667286 on Build GRCh38
rs1041649176 Chr.17: 63666940-63666940 on Build GRCh38
rs761104061 Chr.17: 63666979-63666979 on Build GRCh38
rs759442484 Chr.17: 63667012-63667012 on Build GRCh38
rs1001496507 Chr.17: 63667186-63667186 on Build GRCh38
rs554510109 Chr.17: 63667138-63667138 on Build GRCh38
rs769988237 Chr.17: 63667078-63667078 on Build GRCh38
rs1034221113 Chr.17: 63667207-63667207 on Build GRCh38
rs895701654 Chr.17: 63667265-63667265 on Build GRCh38
rs768845358 Chr.17: 63667039-63667039 on Build GRCh38
rs374801462 Chr.17: 63666985-63666985 on Build GRCh38
rs201709115 Chr.17: 63667013-63667013 on Build GRCh38
rs559388758 Chr.17: 63666893-63666893 on Build GRCh38
rs139576613 Chr.17: 63667000-63667000 on Build GRCh38
rs773535213 Chr.17: 63667009-63667009 on Build GRCh38

View Full Product Details