Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101775772-101776353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF8
Entrez Gene ID
2253
Gene Name
fibroblast growth factor 8
Gene Aliases
AIGF, FGF-8, HBGF-8, HH6, KAL6
UniGene
Hs.57710
Gene Chromosome Location
Chr.10: 101770130-101780369 on Build GRCh38
Transcripts
NM_001206389.1 NM_006119.4 NM_033163.3 NM_033164.3 NM_033165.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372486 c.35T>C p.L12P Chr.10: 101775774-101775774 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs909934745 Chr.10: 101776060-101776060 on Build GRCh38
rs571683960 Chr.10: 101776287-101776287 on Build GRCh38
rs776389277 Chr.10: 101775841-101775841 on Build GRCh38
rs1007190769 Chr.10: 101776134-101776134 on Build GRCh38
rs2735432 Chr.10: 101775964-101775964 on Build GRCh38
rs748640762 Chr.10: 101775837-101775838 on Build GRCh38
rs1051604510 Chr.10: 101776103-101776103 on Build GRCh38
rs769442062 Chr.10: 101775857-101775857 on Build GRCh38
rs934746020 Chr.10: 101775806-101775806 on Build GRCh38
rs933227663 Chr.10: 101776096-101776096 on Build GRCh38
rs889032356 Chr.10: 101776132-101776132 on Build GRCh38
rs1036153984 Chr.10: 101776086-101776086 on Build GRCh38
rs1048444141 Chr.10: 101775797-101775797 on Build GRCh38
rs567972135 Chr.10: 101775826-101775826 on Build GRCh38
rs916160568 Chr.10: 101776001-101776001 on Build GRCh38
rs1021741330 Chr.10: 101776160-101776160 on Build GRCh38
rs977575976 Chr.10: 101775950-101775950 on Build GRCh38
rs879555604 Chr.10: 101776023-101776023 on Build GRCh38
rs779323337 Chr.10: 101775829-101775829 on Build GRCh38
rs150146213 Chr.10: 101775972-101775972 on Build GRCh38
rs746533135 Chr.10: 101775832-101775832 on Build GRCh38
rs1048878615 Chr.10: 101776044-101776044 on Build GRCh38
rs865963475 Chr.10: 101776300-101776300 on Build GRCh38
rs888548294 Chr.10: 101775824-101775824 on Build GRCh38
rs969173777 Chr.10: 101776184-101776199 on Build GRCh38
rs535351281 Chr.10: 101775858-101775858 on Build GRCh38
rs2735430 Chr.10: 101776179-101776179 on Build GRCh38
rs924812178 Chr.10: 101775954-101775954 on Build GRCh38
rs904266819 Chr.10: 101776161-101776161 on Build GRCh38
rs866619890 Chr.10: 101776158-101776158 on Build GRCh38
rs2735429 Chr.10: 101776220-101776220 on Build GRCh38
rs897688568 Chr.10: 101776094-101776094 on Build GRCh38
rs940142999 Chr.10: 101776084-101776084 on Build GRCh38
rs2735431 Chr.10: 101776009-101776009 on Build GRCh38
rs993107382 Chr.10: 101775995-101775995 on Build GRCh38
rs960165703 Chr.10: 101776327-101776327 on Build GRCh38
rs968878417 Chr.10: 101775950-101775950 on Build GRCh38
rs1007655195 Chr.10: 101775929-101775929 on Build GRCh38
rs948726048 Chr.10: 101776012-101776012 on Build GRCh38
rs770331509 Chr.10: 101775838-101775838 on Build GRCh38
rs953104114 Chr.10: 101775859-101775859 on Build GRCh38
rs759091259 Chr.10: 101775844-101775844 on Build GRCh38
rs756187925 Chr.10: 101775935-101775935 on Build GRCh38
rs866323990 Chr.10: 101775924-101775924 on Build GRCh38
rs897739202 Chr.10: 101775841-101775842 on Build GRCh38

View Full Product Details