Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 30812729-30813230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP2CB
Entrez Gene ID
5516
Gene Name
protein phosphatase 2 catalytic subunit beta
Gene Aliases
PP2Abeta, PP2CB
UniGene
Hs.491440
Gene Chromosome Location
Chr.8: 30785610-30812836 on Build GRCh38
Transcripts
NM_001009552.1

SNP Detail

SNP ID Chromosome Location
rs538754443 Chr.8: 30812831-30812831 on Build GRCh38
rs1008772861 Chr.8: 30813028-30813028 on Build GRCh38
rs765101031 Chr.8: 30812772-30812773 on Build GRCh38
rs1052226810 Chr.8: 30813064-30813064 on Build GRCh38
rs184896622 Chr.8: 30812818-30812818 on Build GRCh38
rs915766268 Chr.8: 30813165-30813165 on Build GRCh38
rs1022258309 Chr.8: 30812916-30812916 on Build GRCh38
rs920880652 Chr.8: 30812968-30812968 on Build GRCh38
rs574466469 Chr.8: 30812935-30812935 on Build GRCh38
rs748885886 Chr.8: 30812859-30812859 on Build GRCh38
rs4733201 Chr.8: 30812874-30812874 on Build GRCh38
rs915731544 Chr.8: 30812892-30812892 on Build GRCh38
rs566589527 Chr.8: 30812824-30812824 on Build GRCh38
rs748693871 Chr.8: 30813112-30813112 on Build GRCh38
rs1019065791 Chr.8: 30812842-30812842 on Build GRCh38
rs576972658 Chr.8: 30813144-30813144 on Build GRCh38
rs571016872 Chr.8: 30813078-30813078 on Build GRCh38
rs990412344 Chr.8: 30812890-30812890 on Build GRCh38
rs1037295989 Chr.8: 30812814-30812814 on Build GRCh38
rs1010829166 Chr.8: 30812898-30812898 on Build GRCh38
rs954471229 Chr.8: 30812837-30812837 on Build GRCh38
rs758452711 Chr.8: 30812894-30812894 on Build GRCh38
rs554885468 Chr.8: 30812918-30812918 on Build GRCh38
rs992982332 Chr.8: 30812942-30812942 on Build GRCh38
rs376352755 Chr.8: 30813068-30813068 on Build GRCh38
rs908058644 Chr.8: 30812946-30812946 on Build GRCh38
rs558418245 Chr.8: 30812851-30812851 on Build GRCh38
rs773224209 Chr.8: 30813198-30813198 on Build GRCh38
rs966923302 Chr.8: 30813133-30813133 on Build GRCh38
rs965705046 Chr.8: 30812844-30812844 on Build GRCh38
rs1057489267 Chr.8: 30812772-30812772 on Build GRCh38
rs757716197 Chr.8: 30812833-30812833 on Build GRCh38
rs1045645483 Chr.8: 30812926-30812926 on Build GRCh38
rs530063492 Chr.8: 30812794-30812794 on Build GRCh38
rs928576154 Chr.8: 30813188-30813188 on Build GRCh38
rs779763971 Chr.8: 30812854-30812854 on Build GRCh38
rs767302592 Chr.8: 30812787-30812787 on Build GRCh38
rs772610836 Chr.8: 30813192-30813192 on Build GRCh38
rs759047850 Chr.8: 30812767-30812767 on Build GRCh38
rs940907063 Chr.8: 30812950-30812950 on Build GRCh38
rs754240764 Chr.8: 30812827-30812827 on Build GRCh38
rs940959681 Chr.8: 30812798-30812798 on Build GRCh38
rs898610644 Chr.8: 30812964-30812964 on Build GRCh38
rs989906182 Chr.8: 30813156-30813156 on Build GRCh38
rs890233138 Chr.8: 30812858-30812858 on Build GRCh38
rs115550736 Chr.8: 30812981-30812981 on Build GRCh38
rs1036972530 Chr.8: 30812939-30812939 on Build GRCh38
rs1054260685 Chr.8: 30812755-30812755 on Build GRCh38
rs898559840 Chr.8: 30812809-30812809 on Build GRCh38
rs1010776986 Chr.8: 30813086-30813086 on Build GRCh38
rs1031322608 Chr.8: 30813143-30813143 on Build GRCh38
rs968696990 Chr.8: 30812917-30812917 on Build GRCh38
rs188884972 Chr.8: 30812887-30812887 on Build GRCh38
rs907072660 Chr.8: 30812928-30812928 on Build GRCh38
rs931655712 Chr.8: 30812978-30812978 on Build GRCh38
rs1037915755 Chr.8: 30812958-30812958 on Build GRCh38
rs981655219 Chr.8: 30813173-30813173 on Build GRCh38
rs889959819 Chr.8: 30813023-30813023 on Build GRCh38
rs545887850 Chr.8: 30813205-30813205 on Build GRCh38
rs1015642151 Chr.8: 30812803-30812803 on Build GRCh38
rs1022206131 Chr.8: 30813089-30813089 on Build GRCh38
rs948678640 Chr.8: 30812909-30812909 on Build GRCh38
rs552819831 Chr.8: 30812841-30812841 on Build GRCh38
rs927382149 Chr.8: 30812928-30812929 on Build GRCh38
rs764619945 Chr.8: 30812802-30812802 on Build GRCh38
rs977103522 Chr.8: 30813138-30813138 on Build GRCh38
rs140508959 Chr.8: 30812985-30812985 on Build GRCh38
rs969917682 Chr.8: 30813171-30813171 on Build GRCh38
rs545152285 Chr.8: 30812797-30812797 on Build GRCh38
rs1019873722 Chr.8: 30813129-30813129 on Build GRCh38
rs995652279 Chr.8: 30812819-30812819 on Build GRCh38
rs948644428 Chr.8: 30812779-30812779 on Build GRCh38
rs935827534 Chr.8: 30812749-30812749 on Build GRCh38

View Full Product Details