Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 34321228-34321614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCL8
Entrez Gene ID
6355
Gene Name
C-C motif chemokine ligand 8
Gene Aliases
HC14, MCP-2, MCP2, SCYA10, SCYA8
UniGene
Hs.271387
Gene Chromosome Location
Chr.17: 34319047-34321402 on Build GRCh38
Transcripts
NM_005623.2

SNP Detail

SNP ID Chromosome Location
rs991965502 Chr.17: 34321318-34321318 on Build GRCh38
rs947007909 Chr.17: 34321302-34321302 on Build GRCh38
rs777114497 Chr.17: 34321548-34321548 on Build GRCh38
rs913335471 Chr.17: 34321579-34321579 on Build GRCh38
rs200284380 Chr.17: 34321390-34321390 on Build GRCh38
rs202219411 Chr.17: 34321381-34321381 on Build GRCh38
rs200421610 Chr.17: 34321383-34321383 on Build GRCh38
rs1035474901 Chr.17: 34321349-34321349 on Build GRCh38
rs751400385 Chr.17: 34321594-34321594 on Build GRCh38
rs972554158 Chr.17: 34321495-34321495 on Build GRCh38
rs944079986 Chr.17: 34321535-34321535 on Build GRCh38
rs1024704109 Chr.17: 34321480-34321480 on Build GRCh38
rs201472837 Chr.17: 34321385-34321385 on Build GRCh38
rs938414142 Chr.17: 34321481-34321481 on Build GRCh38
rs115605759 Chr.17: 34321520-34321520 on Build GRCh38
rs749975872 Chr.17: 34321485-34321485 on Build GRCh38
rs1044447305 Chr.17: 34321449-34321449 on Build GRCh38
rs201937316 Chr.17: 34321341-34321341 on Build GRCh38
rs201413984 Chr.17: 34321360-34321360 on Build GRCh38
rs931285432 Chr.17: 34321376-34321377 on Build GRCh38
rs1013744853 Chr.17: 34321459-34321459 on Build GRCh38
rs200430361 Chr.17: 34321342-34321342 on Build GRCh38
rs557315235 Chr.17: 34321458-34321458 on Build GRCh38
rs201092550 Chr.17: 34321283-34321283 on Build GRCh38
rs184120436 Chr.17: 34321339-34321339 on Build GRCh38
rs975144737 Chr.17: 34321382-34321382 on Build GRCh38
rs959391751 Chr.17: 34321317-34321317 on Build GRCh38
rs948368954 Chr.17: 34321429-34321429 on Build GRCh38
rs377385560 Chr.17: 34321359-34321359 on Build GRCh38
rs202171098 Chr.17: 34321327-34321327 on Build GRCh38
rs1024759930 Chr.17: 34321332-34321332 on Build GRCh38
rs201121563 Chr.17: 34321391-34321391 on Build GRCh38
rs574078766 Chr.17: 34321511-34321511 on Build GRCh38
rs1016108892 Chr.17: 34321377-34321377 on Build GRCh38
rs1037534498 Chr.17: 34321522-34321522 on Build GRCh38
rs199540506 Chr.17: 34321410-34321410 on Build GRCh38
rs944120026 Chr.17: 34321383-34321384 on Build GRCh38
rs200096521 Chr.17: 34321337-34321337 on Build GRCh38
rs199734704 Chr.17: 34321389-34321389 on Build GRCh38
rs1033765949 Chr.17: 34321439-34321439 on Build GRCh38
rs72825091 Chr.17: 34321412-34321412 on Build GRCh38
rs543971917 Chr.17: 34321448-34321448 on Build GRCh38
rs919861964 Chr.17: 34321367-34321367 on Build GRCh38
rs1004693842 Chr.17: 34321365-34321365 on Build GRCh38
rs542761104 Chr.17: 34321512-34321512 on Build GRCh38
rs542628552 Chr.17: 34321358-34321359 on Build GRCh38
rs927009015 Chr.17: 34321451-34321451 on Build GRCh38
rs1056785781 Chr.17: 34321508-34321508 on Build GRCh38
rs377230927 Chr.17: 34321569-34321569 on Build GRCh38
rs924039199 Chr.17: 34321386-34321386 on Build GRCh38
rs954943390 Chr.17: 34321384-34321384 on Build GRCh38
rs369968443 Chr.17: 34321347-34321347 on Build GRCh38
rs558500621 Chr.17: 34321404-34321404 on Build GRCh38
rs1044070487 Chr.17: 34321304-34321304 on Build GRCh38
rs752673742 Chr.17: 34321477-34321477 on Build GRCh38
rs528482422 Chr.17: 34321551-34321551 on Build GRCh38

View Full Product Details