Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159315094-159315651 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
UniGene
Hs.674
Gene Chromosome Location
Chr.5: 159314783-159330473 on Build GRCh38
Transcripts
NM_002187.2

SNP Detail

SNP ID Chromosome Location
rs531981828 Chr.5: 159315488-159315488 on Build GRCh38
rs563941424 Chr.5: 159315261-159315261 on Build GRCh38
rs750061605 Chr.5: 159315173-159315173 on Build GRCh38
rs894156001 Chr.5: 159315403-159315403 on Build GRCh38
rs569548120 Chr.5: 159315349-159315349 on Build GRCh38
rs376593057 Chr.5: 159315382-159315382 on Build GRCh38
rs1002901398 Chr.5: 159315383-159315383 on Build GRCh38
rs977306476 Chr.5: 159315200-159315200 on Build GRCh38
rs146117745 Chr.5: 159315543-159315543 on Build GRCh38
rs1032884619 Chr.5: 159315525-159315525 on Build GRCh38
rs545035344 Chr.5: 159315131-159315131 on Build GRCh38
rs528573168 Chr.5: 159315604-159315604 on Build GRCh38
rs991829601 Chr.5: 159315198-159315198 on Build GRCh38
rs746568584 Chr.5: 159315350-159315350 on Build GRCh38
rs921300403 Chr.5: 159315237-159315237 on Build GRCh38
rs867827064 Chr.5: 159315603-159315603 on Build GRCh38
rs41454346 Chr.5: 159315385-159315385 on Build GRCh38
rs898666469 Chr.5: 159315441-159315441 on Build GRCh38
rs886060354 Chr.5: 159315199-159315200 on Build GRCh38
rs960690405 Chr.5: 159315193-159315193 on Build GRCh38
rs56409614 Chr.5: 159315218-159315218 on Build GRCh38
rs144366632 Chr.5: 159315413-159315413 on Build GRCh38
rs1024001080 Chr.5: 159315201-159315201 on Build GRCh38
rs994334879 Chr.5: 159315458-159315458 on Build GRCh38
rs778671255 Chr.5: 159315597-159315597 on Build GRCh38
rs185933681 Chr.5: 159315121-159315121 on Build GRCh38
rs774743402 Chr.5: 159315427-159315427 on Build GRCh38
rs957266336 Chr.5: 159315537-159315537 on Build GRCh38
rs56390703 Chr.5: 159315417-159315417 on Build GRCh38
rs1025432091 Chr.5: 159315468-159315468 on Build GRCh38
rs977256249 Chr.5: 159315509-159315509 on Build GRCh38
rs975436197 Chr.5: 159315230-159315230 on Build GRCh38
rs935128765 Chr.5: 159315246-159315246 on Build GRCh38
rs41292468 Chr.5: 159315355-159315355 on Build GRCh38
rs139698981 Chr.5: 159315402-159315402 on Build GRCh38
rs765892969 Chr.5: 159315307-159315307 on Build GRCh38
rs969312431 Chr.5: 159315203-159315203 on Build GRCh38
rs887254958 Chr.5: 159315275-159315275 on Build GRCh38
rs139879928 Chr.5: 159315571-159315571 on Build GRCh38
rs369157824 Chr.5: 159315405-159315405 on Build GRCh38
rs917070852 Chr.5: 159315159-159315159 on Build GRCh38
rs34324765 Chr.5: 159315200-159315200 on Build GRCh38
rs1051641377 Chr.5: 159315274-159315274 on Build GRCh38
rs868227011 Chr.5: 159315615-159315615 on Build GRCh38
rs534106315 Chr.5: 159315453-159315453 on Build GRCh38
rs967170199 Chr.5: 159315470-159315470 on Build GRCh38
rs975766918 Chr.5: 159315548-159315548 on Build GRCh38
rs934079591 Chr.5: 159315245-159315245 on Build GRCh38
rs55876812 Chr.5: 159315170-159315170 on Build GRCh38
rs549706242 Chr.5: 159315321-159315321 on Build GRCh38
rs993925677 Chr.5: 159315140-159315140 on Build GRCh38
rs979732879 Chr.5: 159315207-159315207 on Build GRCh38
rs767262445 Chr.5: 159315157-159315157 on Build GRCh38
rs762130179 Chr.5: 159315473-159315473 on Build GRCh38
rs3213114 Chr.5: 159315581-159315581 on Build GRCh38
rs1013691100 Chr.5: 159315556-159315556 on Build GRCh38

View Full Product Details