Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180603265-180603417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311238 c.3987C>T p.G1329G Chr.5: 180603297-180603297 on Build GRCh38
4992320 c.3944C>T p.S1315F Chr.5: 180603340-180603340 on Build GRCh38
3761168 c.3971G>T p.R1324L Chr.5: 180603313-180603313 on Build GRCh38
6909969 c.3970C>T p.R1324W Chr.5: 180603314-180603314 on Build GRCh38
6963206 c.3950G>A p.G1317E Chr.5: 180603334-180603334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765704568 Chr.5: 180603299-180603299 on Build GRCh38
rs200963530 Chr.5: 180603291-180603291 on Build GRCh38
rs766508041 Chr.5: 180603347-180603347 on Build GRCh38
rs199520663 Chr.5: 180603388-180603388 on Build GRCh38
rs185808242 Chr.5: 180603346-180603346 on Build GRCh38
rs759044121 Chr.5: 180603283-180603283 on Build GRCh38
rs146806202 Chr.5: 180603376-180603376 on Build GRCh38
rs762359092 Chr.5: 180603296-180603296 on Build GRCh38
rs150279372 Chr.5: 180603326-180603326 on Build GRCh38
rs755210727 Chr.5: 180603312-180603312 on Build GRCh38
rs745546499 Chr.5: 180603323-180603323 on Build GRCh38
rs758489407 Chr.5: 180603305-180603305 on Build GRCh38
rs200392269 Chr.5: 180603320-180603320 on Build GRCh38
rs751544194 Chr.5: 180603351-180603351 on Build GRCh38
rs773431033 Chr.5: 180603320-180603322 on Build GRCh38
rs142322370 Chr.5: 180603329-180603329 on Build GRCh38
rs145806915 Chr.5: 180603328-180603328 on Build GRCh38
rs565053503 Chr.5: 180603290-180603290 on Build GRCh38
rs79620092 Chr.5: 180603322-180603322 on Build GRCh38
rs307821 Chr.5: 180603313-180603313 on Build GRCh38
rs772530628 Chr.5: 180603307-180603308 on Build GRCh38
rs752599369 Chr.5: 180603394-180603394 on Build GRCh38
rs530797555 Chr.5: 180603321-180603321 on Build GRCh38
rs766308772 Chr.5: 180603307-180603307 on Build GRCh38
rs747528649 Chr.5: 180603304-180603304 on Build GRCh38
rs1021827251 Chr.5: 180603288-180603288 on Build GRCh38
rs751807653 Chr.5: 180603311-180603311 on Build GRCh38
rs115824945 Chr.5: 180603325-180603325 on Build GRCh38
rs773779407 Chr.5: 180603335-180603335 on Build GRCh38
rs777755208 Chr.5: 180603314-180603314 on Build GRCh38

View Full Product Details