Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41220601-41220766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXL
Entrez Gene ID
558
Gene Name
AXL receptor tyrosine kinase
Gene Aliases
ARK, JTK11, Tyro7, UFO
UniGene
Hs.590970
Gene Chromosome Location
Chr.19: 41219199-41261767 on Build GRCh38
Transcripts
NM_001699.5 NM_021913.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5021040 c.153G>A p.T51T Chr.19: 41220703-41220703 on Build GRCh38
259589 c.128A>C p.N43T Chr.19: 41220678-41220678 on Build GRCh38
7201261 c.166T>G p.C56G Chr.19: 41220716-41220716 on Build GRCh38
3226719 c.148C>T p.L50F Chr.19: 41220698-41220698 on Build GRCh38
3226725 c.207G>A p.W69* Chr.19: 41220757-41220757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs995493128 Chr.19: 41220741-41220741 on Build GRCh38
rs749915611 Chr.19: 41220627-41220627 on Build GRCh38
rs765631388 Chr.19: 41220710-41220710 on Build GRCh38
rs749268016 Chr.19: 41220672-41220672 on Build GRCh38
rs777529927 Chr.19: 41220670-41220670 on Build GRCh38
rs146598679 Chr.19: 41220745-41220745 on Build GRCh38
rs550455192 Chr.19: 41220739-41220739 on Build GRCh38
rs201764420 Chr.19: 41220640-41220640 on Build GRCh38
rs770570685 Chr.19: 41220680-41220680 on Build GRCh38
rs536724919 Chr.19: 41220657-41220657 on Build GRCh38
rs755527465 Chr.19: 41220744-41220744 on Build GRCh38
rs575141970 Chr.19: 41220737-41220737 on Build GRCh38
rs55767963 Chr.19: 41220703-41220703 on Build GRCh38
rs754682210 Chr.19: 41220641-41220641 on Build GRCh38
rs369684238 Chr.19: 41220698-41220698 on Build GRCh38
rs747638130 Chr.19: 41220663-41220663 on Build GRCh38
rs201081309 Chr.19: 41220654-41220654 on Build GRCh38
rs775741490 Chr.19: 41220696-41220696 on Build GRCh38
rs774132867 Chr.19: 41220692-41220692 on Build GRCh38
rs200868176 Chr.19: 41220722-41220722 on Build GRCh38
rs144418422 Chr.19: 41220728-41220728 on Build GRCh38
rs781049505 Chr.19: 41220662-41220662 on Build GRCh38
rs568179015 Chr.19: 41220702-41220702 on Build GRCh38
rs755688015 Chr.19: 41220664-41220664 on Build GRCh38
rs200598880 Chr.19: 41220693-41220693 on Build GRCh38
rs750804190 Chr.19: 41220714-41220714 on Build GRCh38
rs891614619 Chr.19: 41220639-41220639 on Build GRCh38
rs374467939 Chr.19: 41220706-41220706 on Build GRCh38
rs765658267 Chr.19: 41220628-41220628 on Build GRCh38

View Full Product Details