Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180611389-180611525 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_002020.4 NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009264.1 XM_017009265.1 XM_017009266.1 XM_017009267.1 XM_017009268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6958237 c.3538G>A p.E1180K Chr.5: 180611479-180611479 on Build GRCh38
6665309 c.3595T>G p.F1199V Chr.5: 180611422-180611422 on Build GRCh38
6929170 c.3599C>T p.S1200L Chr.5: 180611418-180611418 on Build GRCh38
6917937 c.3563C>T p.P1188L Chr.5: 180611454-180611454 on Build GRCh38
6239885 c.3582A>T p.S1194S Chr.5: 180611435-180611435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779186080 Chr.5: 180611417-180611417 on Build GRCh38
rs938103588 Chr.5: 180611440-180611440 on Build GRCh38
rs761211590 Chr.5: 180611484-180611484 on Build GRCh38
rs753884372 Chr.5: 180611413-180611413 on Build GRCh38
rs750692929 Chr.5: 180611495-180611495 on Build GRCh38
rs530215661 Chr.5: 180611485-180611485 on Build GRCh38
rs530977029 Chr.5: 180611418-180611418 on Build GRCh38
rs753978823 Chr.5: 180611488-180611488 on Build GRCh38
rs1005859117 Chr.5: 180611467-180611467 on Build GRCh38
rs768688678 Chr.5: 180611439-180611439 on Build GRCh38
rs888048178 Chr.5: 180611465-180611465 on Build GRCh38
rs779959568 Chr.5: 180611428-180611428 on Build GRCh38
rs757374652 Chr.5: 180611491-180611491 on Build GRCh38
rs139544491 Chr.5: 180611463-180611463 on Build GRCh38
rs199984494 Chr.5: 180611453-180611453 on Build GRCh38
rs765166667 Chr.5: 180611494-180611494 on Build GRCh38
rs762445498 Chr.5: 180611461-180611461 on Build GRCh38
rs763760623 Chr.5: 180611467-180611469 on Build GRCh38
rs744282 Chr.5: 180611452-180611452 on Build GRCh38
rs758221491 Chr.5: 180611425-180611425 on Build GRCh38
rs772928672 Chr.5: 180611454-180611454 on Build GRCh38
rs757488795 Chr.5: 180611416-180611416 on Build GRCh38
rs370304760 Chr.5: 180611451-180611451 on Build GRCh38
rs758503543 Chr.5: 180611497-180611497 on Build GRCh38
rs775810969 Chr.5: 180611471-180611471 on Build GRCh38
rs780039630 Chr.5: 180611506-180611506 on Build GRCh38

View Full Product Details