Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180609883-180610048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_002020.4 NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009264.1 XM_017009265.1 XM_017009266.1 XM_017009267.1 XM_017009268.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5403230 c.3729G>A p.G1243G Chr.5: 180609983-180609983 on Build GRCh38
5608888 c.3720G>A p.L1240L Chr.5: 180609992-180609992 on Build GRCh38
7116465 c.3696C>T p.N1232N Chr.5: 180610016-180610016 on Build GRCh38
482649 c.3731C>T p.A1244V Chr.5: 180609981-180609981 on Build GRCh38
6915064 c.3766G>C p.E1256Q Chr.5: 180609946-180609946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746207295 Chr.5: 180609934-180609934 on Build GRCh38
rs747495466 Chr.5: 180609950-180609950 on Build GRCh38
rs201542981 Chr.5: 180609951-180609951 on Build GRCh38
rs759044742 Chr.5: 180609935-180609935 on Build GRCh38
rs769978743 Chr.5: 180609962-180609962 on Build GRCh38
rs971065703 Chr.5: 180609969-180609969 on Build GRCh38
rs370452038 Chr.5: 180609998-180609998 on Build GRCh38
rs755743097 Chr.5: 180609923-180609923 on Build GRCh38
rs371214562 Chr.5: 180609907-180609907 on Build GRCh38
rs142649193 Chr.5: 180609959-180609959 on Build GRCh38
rs773030985 Chr.5: 180609970-180609970 on Build GRCh38
rs780696184 Chr.5: 180609937-180609937 on Build GRCh38
rs376656307 Chr.5: 180610021-180610021 on Build GRCh38
rs752325680 Chr.5: 180609920-180609920 on Build GRCh38
rs750704776 Chr.5: 180609977-180609977 on Build GRCh38
rs200178139 Chr.5: 180609972-180609972 on Build GRCh38
rs1037109406 Chr.5: 180610001-180610001 on Build GRCh38
rs766960409 Chr.5: 180610000-180610000 on Build GRCh38
rs770911540 Chr.5: 180609996-180609996 on Build GRCh38
rs762732020 Chr.5: 180609973-180609973 on Build GRCh38
rs999488864 Chr.5: 180609933-180609933 on Build GRCh38
rs375662657 Chr.5: 180609924-180609924 on Build GRCh38

View Full Product Details