Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111975256-111975529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MERTK
Entrez Gene ID
10461
Gene Name
MER proto-oncogene, tyrosine kinase
Gene Aliases
MER, RP38, Tyro12, c-Eyk, c-mer
UniGene
Hs.306178
Gene Chromosome Location
Chr.2: 111898479-112039946 on Build GRCh38
Transcripts
NM_006343.2 XM_005263565.4 XM_005263568.4 XM_011510490.2 XM_017003164.1 XM_017003165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4403026 c.961-1G>A p.? Chr.2: 111975288-111975288 on Build GRCh38
6257639 c.1072G>T p.G358C Chr.2: 111975400-111975400 on Build GRCh38
6261023 c.1045C>T p.L349L Chr.2: 111975373-111975373 on Build GRCh38
6262155 c.1047G>T p.L349L Chr.2: 111975375-111975375 on Build GRCh38
6711509 c.1017C>A p.A339A Chr.2: 111975345-111975345 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752126753 Chr.2: 111975393-111975393 on Build GRCh38
rs371465555 Chr.2: 111975283-111975283 on Build GRCh38
rs372879468 Chr.2: 111975496-111975496 on Build GRCh38
rs760569673 Chr.2: 111975344-111975344 on Build GRCh38
rs778299669 Chr.2: 111975459-111975459 on Build GRCh38
rs200167805 Chr.2: 111975332-111975332 on Build GRCh38
rs759172948 Chr.2: 111975313-111975313 on Build GRCh38
rs771431781 Chr.2: 111975481-111975482 on Build GRCh38
rs745461381 Chr.2: 111975461-111975461 on Build GRCh38
rs868566811 Chr.2: 111975418-111975418 on Build GRCh38
rs960364215 Chr.2: 111975501-111975501 on Build GRCh38
rs377295767 Chr.2: 111975502-111975502 on Build GRCh38
rs151145412 Chr.2: 111975412-111975412 on Build GRCh38
rs1019443525 Chr.2: 111975295-111975295 on Build GRCh38
rs747489065 Chr.2: 111975282-111975282 on Build GRCh38
rs953412461 Chr.2: 111975442-111975442 on Build GRCh38
rs142060697 Chr.2: 111975386-111975386 on Build GRCh38
rs781756280 Chr.2: 111975421-111975421 on Build GRCh38
rs561718480 Chr.2: 111975305-111975305 on Build GRCh38
rs892521266 Chr.2: 111975349-111975349 on Build GRCh38
rs777376015 Chr.2: 111975287-111975287 on Build GRCh38
rs756814656 Chr.2: 111975434-111975434 on Build GRCh38
rs773169183 Chr.2: 111975499-111975499 on Build GRCh38
rs967891555 Chr.2: 111975417-111975417 on Build GRCh38
rs748786937 Chr.2: 111975304-111975304 on Build GRCh38
rs886054757 Chr.2: 111975324-111975324 on Build GRCh38
rs113052219 Chr.2: 111975400-111975400 on Build GRCh38
rs761792381 Chr.2: 111975485-111975485 on Build GRCh38
rs746786777 Chr.2: 111975468-111975468 on Build GRCh38
rs777350533 Chr.2: 111975312-111975313 on Build GRCh38
rs748733403 Chr.2: 111975423-111975423 on Build GRCh38
rs112257146 Chr.2: 111975306-111975306 on Build GRCh38
rs776597215 Chr.2: 111975371-111975371 on Build GRCh38
rs761847323 Chr.2: 111975381-111975381 on Build GRCh38
rs768477896 Chr.2: 111975470-111975470 on Build GRCh38
rs991749351 Chr.2: 111975503-111975503 on Build GRCh38
rs764044468 Chr.2: 111975362-111975362 on Build GRCh38
rs1019043513 Chr.2: 111975324-111975324 on Build GRCh38
rs758562603 Chr.2: 111975389-111975389 on Build GRCh38
rs776542197 Chr.2: 111975471-111975471 on Build GRCh38
rs748817458 Chr.2: 111975369-111975370 on Build GRCh38
rs199942623 Chr.2: 111975382-111975382 on Build GRCh38
rs985189779 Chr.2: 111975480-111975480 on Build GRCh38
rs150487517 Chr.2: 111975462-111975462 on Build GRCh38
rs369339515 Chr.2: 111975455-111975455 on Build GRCh38
rs775153333 Chr.2: 111975338-111975338 on Build GRCh38
rs34943572 Chr.2: 111975314-111975314 on Build GRCh38

View Full Product Details