Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63645933-63646206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K3
Entrez Gene ID
4215
Gene Name
mitogen-activated protein kinase kinase kinase 3
Gene Aliases
MAPKKK3, MEKK3
UniGene
Hs.29282
Gene Chromosome Location
Chr.17: 63622209-63696310 on Build GRCh38
Transcripts
NM_002401.3 NM_203351.1 XM_005257376.3 XM_005257377.3 XM_005257378.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3421775 c.230G>A p.R77K Chr.17: 63646044-63646044 on Build GRCh38
1750225 c.133G>A p.V45I Chr.17: 63646040-63646040 on Build GRCh38
3520770 c.259C>G p.R87G Chr.17: 63646073-63646073 on Build GRCh38
3421776 c.137G>A p.R46K Chr.17: 63646044-63646044 on Build GRCh38
3764620 c.132C>T p.D44D Chr.17: 63646039-63646039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775102842 Chr.17: 63646090-63646090 on Build GRCh38
rs149314666 Chr.17: 63646064-63646064 on Build GRCh38
rs758870331 Chr.17: 63646116-63646116 on Build GRCh38
rs779324350 Chr.17: 63646020-63646020 on Build GRCh38
rs115334804 Chr.17: 63646004-63646004 on Build GRCh38
rs767043506 Chr.17: 63645985-63645985 on Build GRCh38
rs999233996 Chr.17: 63646161-63646161 on Build GRCh38
rs771488402 Chr.17: 63646078-63646078 on Build GRCh38
rs766476291 Chr.17: 63646117-63646117 on Build GRCh38
rs375250733 Chr.17: 63646063-63646063 on Build GRCh38
rs948635882 Chr.17: 63645963-63645963 on Build GRCh38
rs750933927 Chr.17: 63646026-63646026 on Build GRCh38
rs757697531 Chr.17: 63646011-63646012 on Build GRCh38
rs1001024067 Chr.17: 63646040-63646040 on Build GRCh38
rs750905700 Chr.17: 63646106-63646106 on Build GRCh38
rs755299577 Chr.17: 63646011-63646014 on Build GRCh38
rs754128532 Chr.17: 63645975-63645975 on Build GRCh38
rs372206804 Chr.17: 63646037-63646037 on Build GRCh38
rs754210174 Chr.17: 63646013-63646013 on Build GRCh38
rs776394763 Chr.17: 63646098-63646098 on Build GRCh38
rs76397113 Chr.17: 63646076-63646076 on Build GRCh38
rs770755395 Chr.17: 63646066-63646066 on Build GRCh38
rs748388819 Chr.17: 63646125-63646125 on Build GRCh38
rs765617563 Chr.17: 63646104-63646104 on Build GRCh38
rs752897281 Chr.17: 63646090-63646091 on Build GRCh38
rs369281595 Chr.17: 63646103-63646103 on Build GRCh38
rs755152020 Chr.17: 63646054-63646054 on Build GRCh38
rs763892428 Chr.17: 63646096-63646096 on Build GRCh38
rs184639184 Chr.17: 63646070-63646070 on Build GRCh38
rs745762268 Chr.17: 63646071-63646071 on Build GRCh38
rs535724632 Chr.17: 63645969-63645969 on Build GRCh38
rs756402054 Chr.17: 63646095-63646102 on Build GRCh38
rs758462712 Chr.17: 63646027-63646027 on Build GRCh38
rs747235726 Chr.17: 63646050-63646050 on Build GRCh38
rs752413223 Chr.17: 63645999-63645999 on Build GRCh38
rs533949737 Chr.17: 63646007-63646007 on Build GRCh38
rs760427972 Chr.17: 63646093-63646093 on Build GRCh38
rs777707925 Chr.17: 63646125-63646126 on Build GRCh38
rs372488037 Chr.17: 63646122-63646122 on Build GRCh38
rs757622671 Chr.17: 63646017-63646017 on Build GRCh38
rs759035815 Chr.17: 63645983-63645983 on Build GRCh38
rs549035028 Chr.17: 63645978-63645978 on Build GRCh38
rs751588401 Chr.17: 63646119-63646119 on Build GRCh38

View Full Product Details