Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159320265-159320494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
UniGene
Hs.674
Gene Chromosome Location
Chr.5: 159314783-159330473 on Build GRCh38
Transcripts
NM_002187.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7090535 c.542G>T p.R181I Chr.5: 159320461-159320461 on Build GRCh38
7107541 c.593C>T p.A198V Chr.5: 159320410-159320410 on Build GRCh38
7258035 c.688A>G p.R230G Chr.5: 159320315-159320315 on Build GRCh38
5402920 c.681C>T p.F227F Chr.5: 159320322-159320322 on Build GRCh38
6255589 c.630C>A p.V210V Chr.5: 159320373-159320373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748215576 Chr.5: 159320343-159320343 on Build GRCh38
rs756166931 Chr.5: 159320472-159320472 on Build GRCh38
rs375159171 Chr.5: 159320467-159320467 on Build GRCh38
rs905944344 Chr.5: 159320376-159320376 on Build GRCh38
rs138127918 Chr.5: 159320380-159320380 on Build GRCh38
rs763525702 Chr.5: 159320405-159320405 on Build GRCh38
rs923137937 Chr.5: 159320316-159320316 on Build GRCh38
rs777742449 Chr.5: 159320474-159320474 on Build GRCh38
rs764913762 Chr.5: 159320434-159320434 on Build GRCh38
rs79446920 Chr.5: 159320360-159320360 on Build GRCh38
rs769766217 Chr.5: 159320352-159320352 on Build GRCh38
rs142400793 Chr.5: 159320448-159320448 on Build GRCh38
rs10045130 Chr.5: 159320459-159320459 on Build GRCh38
rs771282654 Chr.5: 159320362-159320362 on Build GRCh38
rs960228537 Chr.5: 159320399-159320399 on Build GRCh38
rs745769395 Chr.5: 159320325-159320325 on Build GRCh38
rs749361927 Chr.5: 159320469-159320469 on Build GRCh38
rs55661460 Chr.5: 159320326-159320326 on Build GRCh38
rs149027358 Chr.5: 159320334-159320334 on Build GRCh38
rs776207541 Chr.5: 159320394-159320394 on Build GRCh38
rs149273360 Chr.5: 159320361-159320361 on Build GRCh38
rs760025343 Chr.5: 159320381-159320381 on Build GRCh38
rs756894865 Chr.5: 159320433-159320433 on Build GRCh38
rs768122910 Chr.5: 159320389-159320389 on Build GRCh38
rs760165869 Chr.5: 159320402-159320402 on Build GRCh38
rs753481336 Chr.5: 159320432-159320432 on Build GRCh38
rs567462873 Chr.5: 159320415-159320415 on Build GRCh38

View Full Product Details