Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180236333-180236584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK9
Entrez Gene ID
5601
Gene Name
mitogen-activated protein kinase 9
Gene Aliases
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK
UniGene
Hs.484371
Gene Chromosome Location
Chr.5: 180233594-180292071 on Build GRCh38
Transcripts
NM_002752.4 NM_139068.2 NM_139069.2 NM_139070.2 XM_005265940.4 XM_006714891.2 XM_017009638.1 XM_017009639.1 XM_017009640.1 XM_017009641.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3012164 c.1175C>T p.S392L Chr.5: 180236484-180236484 on Build GRCh38
1620169 c.1274G>T p.*425L Chr.5: 180236385-180236385 on Build GRCh38
6343844 c.1218C>A p.A406A Chr.5: 180236441-180236441 on Build GRCh38
1620168 c.1274G>T p.*425L Chr.5: 180236385-180236385 on Build GRCh38
6708030 c.1153G>A p.A385T Chr.5: 180236506-180236506 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773149490 Chr.5: 180236468-180236468 on Build GRCh38
rs771276478 Chr.5: 180236419-180236419 on Build GRCh38
rs769536181 Chr.5: 180236553-180236553 on Build GRCh38
rs1027287292 Chr.5: 180236357-180236357 on Build GRCh38
rs115701296 Chr.5: 180236389-180236389 on Build GRCh38
rs746810129 Chr.5: 180236397-180236397 on Build GRCh38
rs768744849 Chr.5: 180236539-180236539 on Build GRCh38
rs767097311 Chr.5: 180236492-180236492 on Build GRCh38
rs561947205 Chr.5: 180236541-180236541 on Build GRCh38
rs776867044 Chr.5: 180236457-180236457 on Build GRCh38
rs754581492 Chr.5: 180236408-180236408 on Build GRCh38
rs991693051 Chr.5: 180236506-180236506 on Build GRCh38
rs780334320 Chr.5: 180236519-180236519 on Build GRCh38
rs765017960 Chr.5: 180236463-180236463 on Build GRCh38
rs779359114 Chr.5: 180236505-180236505 on Build GRCh38
rs200820257 Chr.5: 180236483-180236483 on Build GRCh38
rs749592487 Chr.5: 180236412-180236412 on Build GRCh38
rs548387145 Chr.5: 180236503-180236503 on Build GRCh38
rs115730064 Chr.5: 180236471-180236471 on Build GRCh38
rs371077278 Chr.5: 180236391-180236391 on Build GRCh38
rs114433256 Chr.5: 180236377-180236377 on Build GRCh38
rs576783254 Chr.5: 180236452-180236452 on Build GRCh38
rs980060008 Chr.5: 180236484-180236484 on Build GRCh38
rs747083644 Chr.5: 180236526-180236526 on Build GRCh38
rs772288463 Chr.5: 180236429-180236429 on Build GRCh38
rs996280340 Chr.5: 180236355-180236355 on Build GRCh38
rs751271294 Chr.5: 180236393-180236393 on Build GRCh38
rs747278411 Chr.5: 180236448-180236448 on Build GRCh38
rs765111829 Chr.5: 180236369-180236369 on Build GRCh38
rs200458599 Chr.5: 180236388-180236388 on Build GRCh38
rs149127216 Chr.5: 180236409-180236409 on Build GRCh38
rs748345878 Chr.5: 180236548-180236548 on Build GRCh38
rs763006797 Chr.5: 180236433-180236456 on Build GRCh38
rs917182550 Chr.5: 180236549-180236549 on Build GRCh38
rs754707056 Chr.5: 180236489-180236489 on Build GRCh38
rs527563372 Chr.5: 180236556-180236556 on Build GRCh38
rs531472225 Chr.5: 180236478-180236478 on Build GRCh38
rs746307580 Chr.5: 180236428-180236428 on Build GRCh38
rs147124741 Chr.5: 180236518-180236518 on Build GRCh38
rs114045781 Chr.5: 180236447-180236447 on Build GRCh38
rs761862060 Chr.5: 180236359-180236359 on Build GRCh38
rs374476654 Chr.5: 180236504-180236504 on Build GRCh38
rs115645812 Chr.5: 180236458-180236458 on Build GRCh38
rs115938486 Chr.5: 180236420-180236420 on Build GRCh38
rs763870240 Chr.5: 180236495-180236497 on Build GRCh38
rs968150047 Chr.5: 180236467-180236467 on Build GRCh38

View Full Product Details